Microarray Analysis Instrument
Thermo Fisher Scientific offers a number of Affymetrix instruments for microarray analysis, from integrated platforms to individual scanners, ovens, fluidics stations, and target preparation instruments.
Microarray instruments
The GeneChip™ Scanner 3000 7G System allows you to scan next-generation higher-density arrays, including SNP arrays with up to 900,000 SNPs, tiling arrays for transcription and all-exon arrays.
The GeneChip™ Hybridization Oven 645 provides precise temperature and rotation control to help ensure the successful hybridization of as many as 64 GeneChip-brand cartridges at one time.
The GeneChip™ Fluidics Station 450 is used for the wash and stain operation of Affymetrix™ GeneChip arrays.
An instrument for expression and genotyping experiments that seamlessly integrates hybridization, washing, and imaging in a single instrument to provide automated array processing—whether you are performing basic or applied research
Thermo Fisher Scientific has partnered with Hamilton Robotics to provide you with the latest in ultra-compact and precision-based liquid-handling instrumentation. The NIMBUS Target Preparation Instrument features high-end manufacturing and superior pipetting performance at an affordable price.