Search
Nucleic Acid Extraction for Respiratory Testing |
Testing for respiratory tract infection requires accurate nucleic acid extraction. Achieve high quality results in clinical diagnostic workflow for respiratory infections with the Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit. A workflow that starts with nucleic acid extraction for in vitro diagnostic use can give labs increased confidence in their downstream results. The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit helps ensure high-quality reproducible results since it offers our most advanced formulation. MagMAX Dx Viral/Pathogen NA Isolation Kit is automation compatible with KingFisher Apex Dx for a streamlined modular workflow.
Select between MagMAX Dx Viral/Pathogen NA Isolation Kit and MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit (IVD) as per your diagnostic testing needs.
![]() MagMAX Dx Viral/Pathogen NA Isolation Kit | ![]() MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit (IVD) | |
| What are the major improvements? |
| N/A |
| Regulatory Status | IVD | IVD | CE-IVD |
| Nucleic acid | Viral RNA, viral DNA, gram-positive bacterial nucleic acid and gram-negative bacterial nucleic acid | Viral RNA and viral DNA |
| Sample types | Nasopharyngeal swab, anterior nasal swab, saliva and sputum | Nasopharyngeal swabs and saliva |
| Automation level | Automation ready protocols designed for the KingFisher Apex Dx Purification Instrument | Automation-ready protocols compatible with Thermo Scientific KingFisher Flex purification system |
| Sample input volume | 200–400 µL workflow | 200 or 400 µL |
| Elution volume | Elution volume of 60 µL | Elution volume of 50 µL |
| Time per run | Fast automated procedures that allow for 96 samples to be processed on KingFisher Purification System with ~30 minutes instrument run time | |
| Shelf life | 24 months | |
| Application Compatibility | qPCR workflows | |
| REACH compliancy | REACH Compliant | FSC certified | REACH compliant |
| Cat. No. | A52735 (up to 2,000 preps/kit) A52736 (up to 600 preps/kit) | A48383 (up to 2,000 preps/kit) |
Clinical labs require the ability to consistently recover high-quality nucleic acids for respiratory testing with minimal variability. KingFisher Apex Dx provides consistent recovery of high-quality nucleic acids from well to well and sample to sample. The MagMAX Dx Viral/Pathogen NA Isolation Kit with KingFisher Apex Dx is designed to be part of an automated modular testing workflow that helps maximize nucleic acid yield and delivers precise results.
The workflow includes fast automated procedures that allow for 96 samples to be processed on KingFisher Apex Dx Purification System with ~30 minutes instrument run time. There are two workflow options available to select from with one single script (200 µL workflow or scalable 200–400 µL workflow).
![]()
Receive patient samples
*Swabs collected into viral transport medium (VTM)
Transfer samples to 96 deep well plate
![]() | ![]() |
Extract target nucleic acids with MagMAX Dx Viral/Pathogen NA Isolation Kit on 24-or-96-well plate. Automate nucleic acid purification on KingFisher Apex Dx instrument.
Select panel for either 96 or 384 block plate. Add sample eluent nucleic acids to plate.
Use the eluted samples in downstream qPCR workflows.
![]()
Receive patient samples
*Swabs collected into viral transport medium (VTM)
Transfer samples to 96 deep well plate
![]() | ![]() |
Extract target nucleic acids with MagMAX Dx Viral/Pathogen NA Isolation Kit on 24-or-96-well plate. Automate nucleic acid purification on KingFisher Apex Dx instrument.
Select panel for either 96 or 384 block plate. Add sample eluent nucleic acids to plate.
Use the eluted samples in downstream qPCR workflows.
CV = coefficient variation. For more information, please see IFU.
The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is a magnetic bead technology nucleic acid purification kit intended for the isolation and purification of viral and bacterial nucleic acid from human respiratory biological specimens for qPCR downstream application (see below). The kit is intended for use by qualified and trained clinical laboratory personnel specifically instructed and trained in the techniques of automated magnetic bead purification and in vitro diagnostic procedures.
For In Vitro Diagnostic Use. Regulatory requirements vary by country. Product may not be available in your geographic region.