header-qqq-confident-quant-Banner-1170x250
Confident quantitation with triple quadrupole LC-MS systems
From addressing complexities of molecules and matrices, to ensuring ease-of-use while satisfying regulatory requirements—today’s challenges in targeted quantitation involve more than just measuring concentrations of target analytes. Achieve ultimate confidence in your data quality with a robust, sensitive, reproducible, and reliable targeted quantitation method using our new liquid chromatography mass spectrometry (LC/MS) workflow solutions.
40 year anniversary of Thermo Scientific development and success of triple quadrupole mass spectrometers
For today and what's next
TSQ Altis Quadrupole Mass Spectrometer
Ultimate sensitivity & selectivity to meet tomorrow’s standards today
Learn more
For everyday excellence
TSQ Quantis Quadrupole
Mass Spectrometer
Everyday quantitation workhorse to meet today’s standards
Learn more
For a new level of confidence
TSQ Fortis Triple Quadrupole Mass Spectrometer
Quality data across workflows for users at all levels of expertise
Learn more
Triple quadrupole LC-MS system product comparison
Different applications require different levels of system sensitivity, resolution, and speed. In this table, we compare different TSQ triple quadrupole systems in order to showcase which ones are most relevant to your laboratory's needs. For more information on these systems, click on our Smartnote: Transitioning to Confident Quantitation – in search of a better tomorrow
Triple Quad Product Comparison Fortis Endura Quantis Altis
Sensitivity +++ +++ ++++ +++++
Resolution ++ ++ ++ +++
Scan Speed ++++ +++ ++++ ++++
Applications across multiple matrices
Targeted quantitation in Proteomics No Yes Yes Yes
Peptide and protein quantitation (Biopharma) No Yes Yes Yes
Small molecule quantitation (Pharma, Clinical Research) Yes Yes Yes Yes
Small molecule mixture quantitation (Environmental Safety, Food Safety, Forensic Toxicology, Pharma) Yes Yes Yes Yes
Analysis and targeted quantitation of trace impurities (Impurity Quantitation) Yes Yes Yes Yes
Targeted quantitation in Omics (metabolomics, lipidomics) Yes Yes Yes Yes
Critical Features
MRM for reliable identification Yes Yes Yes Yes
Multiple Source Compatibility Yes Yes Yes Yes
High Resolution SRM: for increased selectivity and sensitivity for analytes or multiple analytes in complex matrices ++ ++ ++ +++
Segmented quadrupoles for increased ion efficiency Yes No Yes Yes
Neutral blocker for increased robustness Yes Yes Yes Yes
Increased dynodes in detector for increased lifetime and better linear dynamic range Yes No Yes Yes
APCI compatible with HESI source Yes No Yes Yes
Polarity Switching: Fast switching with stabilization enable faster quantitation assays for every molecule type + + + +
System Remote Monitoring Yes Yes Yes Yes
Assist with 21 CFR Part 11 Compliance (appropriate software) Yes. With LCQUAN™ Software Yes. With LCQUAN™ Software Yes. With LCQUAN™ Software Yes. With LCQUAN™ Software
Instrument IQ/OQ assist Yes Yes Yes Yes
Thermo Fisher Cloud: Access, process, review data whenever you like, from wherever you are No Yes Yes Yes
Wi-Fi Connectivity: Access your data, Check your assay performance remotely Yes Yes Yes Yes
Size 27x30x33 (in) 27x30x33 (in) 27x30x33 (in) 27x30x33 (in)
IVD Platform No Available in certain countries No No
Compatible separation technologies UltiMate, Vanquish, Multiplexing, Capillary LC, Other vendors UltiMate, Vanquish, Other vendors, IC, Multiplexing, Capillary LC, NanoLC UltiMate, Vanquish, Other vendors, IC, Multiplexing, Capillary LC, NanoLC UltiMate, Vanquish, Other vendors, IC, Multiplexing, Capillary LC, NanoLC
Compatible Software? Xcalibur, TraceFinder, Chromeleon Xcalibur, TraceFinder, Chromeleon Xcalibur, TraceFinder, Chromeleon Xcalibur, TraceFinder, Chromeleon
Library availabilty Yes Yes Yes Yes
Quick link
TSQ Quantum Access MAX Triple Quadrupole Mass Spectrometer
Common applications of triple quadrupole technology
To perform quantitation assays with confidence, you need a liquid chromatography mass spectrometry (LCMS) workflow solution that is relevant to your laboratory’s goals. These workflow solutions are adapted to specific applications, including clinical research, environmental and food safety, forensic toxicology, and pharma/biopharma.
Resources
Brochures
Application notes
Support
Whitepapers
Technical notes