Search
Due to potential health risks, ß-agonists have been banned for use as growth promoters in livestock. As a consequence, global regulatory agencies require highly sensitive and specific analysis methods to ensure that ß-agonist residues are not present in animal-derived foods. The Thermo Scientific™ LC-MS ß-agonist Testing Kit for Livestock and Animal-derived Foods provides the application protocol and products required to meet international regulatory requirements for ß-agonist analysis.
| Complete kit description | Part number | Qty. |
|---|---|---|
| Thermo Scientific™ ß-agonist application starter kit | TS-MKIT0001S | 1 |
| Thermo Scientific™ ß-agonist application one month consumable kit | TS-MKIT0001 | 1 |
ß-Agonist kits can be ordered in complete kits or by individual consumable
| Individual items description | Part number | Qty. |
|---|---|---|
| Thermo Scientific™ HyperSep™ Retain CX SPE cartridges (500mg/3mL; 50/Pk.) | 60107-305 | 1 |
| Thermo Scientific™ Hypersil™ GOLD HPLC column (150 x 2.1mm, 5µm) | 25005-152130 | 1 |
| Compound | LOD (µg/kg) |
|---|---|
| Clenbuterol | 1 |
| Salbutamol | 1 |
| Ractopamine | 3 |