Quickly assess your sample for nucleic acids, proteins, and contaminants
When you understand the quantity and purity of your samples, you can make educated decisions about using them in downstream experiments. The Thermo Scientific NanoDrop spectrophotometer family helps you quickly assess whether your sample contains what you intended in appropriate amounts. Depending on the instrument you choose, the software also can alert you when contaminants are present and identify what they are.
Nucleic acid and protein quantification and purity assessment, the core functions of NanoDrop spectrophotometers, are indispensable for many downstream applications, notably RT-qPCR and quality control. Applications such as bacterial culture growth and kinetics applications are also supported. On our premier models, you can also create and share custom methods or download them from our website.
When unlocked, the Acclaro Pro applications provide enhanced accuracy for analyzing highly concentrated proteins and nucleic acids, such as those encountered in monoclonal antibody production.
Nucleic acid quantification
Nucleic acids have traditionally been quantified by determining the UV absorbance at three analytical wavelengths: 230 nm, 260 nm, and 280 nm. These absorbance measurements allow scientists to measure nucleic acid concentration and have an indication of sample purity. The intensity of their maximum absorbance peak at 260 nm is proportional to nucleic acid concentration.
The advantages of this nucleic acid quantitation method are that it is simple, direct, and consumes only a small volume of your sample. One challenge, however, is its lack of specificity, as any contaminant that absorbs at these wavelengths will lead to inaccuracies in the resultant nucleic acid concentration.
With advances in UV-Vis software, researchers can now selectively quantify nucleic acid absorbance in the presence of chemical and nucleic acid contaminants. Thermo Scientific Acclaro Intelligence Technology software (standard in our premier NanoDrop Ultra spectrophotometers and fluorometers, and NanoDrop Eight spectrophotometers) uses full-spectrum data and advanced algorithms to identify common nucleic acid contaminants and provide corrected nucleic acid concentrations. Acclaro algorithms can detect dsDNA contamination in RNA and RNA contamination in dsDNA. You can also input custom oligo sequences and the software will automatically calculate the factor to apply for nucleic acid quantification.
Preprogrammed nucleic acid applications on the NanoDrop Ultra Spectrophotometers and Fluorometers
Tap to select the application you need from the nucleic acids home screen.
Quantification of labeled nucleic acids
Fluorescently labeled nucleic acids' absorbance can be quantified using the Microarray preconfigured software method that delivers the nucleic acid as well as the label concentration (up to two dyes). This method is available as a preconfigured application on NanoDrop Ultra and NanoDrop Eight instruments.
Contaminant identification
Acclaro Sample Intelligence Technology allows the NanoDrop Ultra Microvolume Spectrophotometers and Fluorometers, and NanoDrop Eight Microvolume Spectrophotometers to provide more information about sample quality by identifying common contaminants and delivering true sample concentrations. Acclaro software incorporates algorithms that rely on a reference library of spectra. The software applies these algorithms to the sample spectrum and makes predictions about the sample's contaminants by using chemometric mathematical principles.
Acclaro Contaminant Identification
Acclaro software has flagged this dsDNA sample (yellow icon) because it is contaminated with protein. The absorbance contribution from the protein (orange) has been subtracted from the original result (total A260, green) to obtain the true, corrected dsDNA concentration in the sample (purple).
The Acclaro reference libraries currently support detection of protein, phenol, guanidine HCl, and RNA in dsDNA samples. They also support detection of protein, phenol, guanidine isothiocyanate, and dsDNA in RNA samples. Finally, they allow detection of DNA in protein samples. Additional contaminants will be added to the libraries in the future.
Contaminant identification gives researchers two important pieces of information. First, it identifies specific contaminants that are likely to be present in the sample. This information can help scientists troubleshoot difficult extractions or purifications and make decisions regarding sample use in downstream experiments. Second, it provides a corrected concentration of the analyte. When setting up downstream reactions such as PCR where DNA concentration is a critical parameter, the corrected concentration will help scientists ensure the success of downstream experiments.
For example, RNA contamination in genomic DNA preparations is a common issue in molecular biology workflows. In traditional spectrophotometry, the presence of copurified RNA can artificially inflate the concentration of DNA. By using the full spectral data and multivariate mathematical algorithms available in the Acclaro software, researchers can identify RNA contamination in a DNA sample and see the corrected DNA concentration results.
An improved Acclaro software is included with the NanoDrop Ultra Spectrophotometers and Fluorometers allowing detection/correction of RNA contamination in DNA and DNA contamination in RNA for Mammalian, Plant, and Bacterial species.
Nucleic acid quantification in RT-qPCR workflows
Reliable RT-qPCR (quantitative reverse transcription PCR) assays demand careful experimental design, extensive quality control (QC), and transparent data analysis.  
Importance of nucleic acid quantification in RT-qPCR QC
Nucleic acid quantification is critical in both enzymatic steps of the RT-qPCR process. In the reverse transcription step, knowing (and normalizing) the amount of RNA going into the reaction helps minimize variability in cDNA production. In the PCR step, you must be certain you have enough cDNA template to determine the genotype of the sample.
The MIQE guidelines—minimum information for publication of quantitative real-time PCR experiments—require that input RNA quantity and purity are reported in publications that use qPCR.
Using NanoDrop spectrophotometers in qPCR workflows
The NanoDrop Ultra spectrophotometers and Fluorometers, and NanoDrop Eight spectrophotometers are well suited to quantify and qualify nucleic acid samples as a quality control step for downstream RT-qPCR assays. These instruments can help prevent failed or unpublishable experiments by providing accurate nucleic acid concentrations and identifying contaminants that may alter experimental results.
Acclaro Sample Intelligence Technology, built into both the NanoDrop Ultra and Eight instruments, identifies the presence of common contaminants in samples and calculates an approximate concentration of the contaminant. Common molecules found in nucleic acid extraction kits can overestimate analyte concentrations or denature qPCR polymerases. In either case, the qPCR experiment can fail. The Acclaro software provides corrected nucleic acid concentrations and identifies contaminants, to help minimize time spent troubleshooting and managing failed experiments.
The NanoDrop Ultra Spectrophotometers and Fluorometers feature an integrated qPCR recipe calculator, removing the need for manual and error-prone calculations of sample volumes for downstream reactions. Utilize the built-in library of kits or add your preferred ones, allowing the software to perform the necessary calculations. The qPCR recipe calculator provides precise preparation instructions based on your sample's concentration.
Fluorescence detection
Understand purity information and quantify highly concentrated samples without dilution. Contaminated samples with the sensitivity and specificity of fluorescence all in one instrument with the NanoDrop Ultra FL and NanoDrop UltraC FL Instruments.
Protein quantification
Scientists can use NanoDrop instruments to quantify the protein content in their sample using either direct or indirect measurements. NanoDrop UV-Vis spectrophotometers and fluorometers support protein sample quantification with applications for direct A280 and A205 measurements in the ultraviolet range and colorimetric assays in the visible range of the spectrum.
Preprogrammed protein applications on the NanoDrop Ultra Spectrophotometers
Tap to select the application you need from the proteins home screen.
Protein A280 is a direct measurement application that calculates protein concentration based directly on the sample absorbance at 280 or 205 nm and the protein-specific extinction coefficient. Direct measurements are a popular choice for researchers because they are simple to perform, require no reagents or standards, and consume very little sample. Proteins, unlike nucleic acids, can exhibit considerable diversity. The Protein A280 application is appropriate for purified proteins that contain residues of tryptophan (Trp) or tyrosine (Tyr) cysteine disulfide bonds (Cys-Cys) and exhibit absorbance at 280 nm.
Colorimetric assays such as BCA, Bradford, Lowry, and Pierce 660 nm, are indirect measurement applications that rely on the generation of a standard curve and a reaction that produces a color change proportional to the amount of protein in the sample. Proteins in complex mixtures such as cell extracts or lysates are best measured using a protein colorimetric assay.
Webinar: Protein sample evaluation using a NanoDrop UV-Vis Spectrophotometer and Fluorometers
Protein sample evaluation is an important step in many protein workflows. This webinar explains how NanoDrop instruments streamline the process, compares the different protein assays available, and reviews best practices for accurate protein quantification.
Choosing the best quantification assay for your protein or peptide sample
The tables below show ranges, descriptions, and considerations for protein quantification methods available on NanoDrop UV-Vis spectrophotometer, along with which methods are provided on which instruments. For more information on some of these assays, see our protein assay selection guide.
Protein methods supported on NanoDrop spectrophotometers
Method
Concentration range for NanoDrop Ultra Spectrophotometers and Fluorometers
Description
Considerations
A280
  • Pedestal: 0.03-820 mg/mL BSA (0.02-402 mg/mL IgG)
  • Cuvette: 0.006–2.38 mg/mL BSA
  • Direct measurement, quick and easy.
  • Best method for pure proteins that contain Trp and Tyr residues.
  • Uses Beer's Law to calculate concentration. For accurate results the correct extinction coefficient must be entered.
  • Does not require standard curve.
  • When measuring protein mixtures use 1 Abs=1 mg/mL “sample type”.
  • Contaminants or buffers that absorb ~280 nm will affect protein concentration calculation.
  • Acclaro Sample Intelligence Technology detects nucleic acid contaminants in protein samples and corrects concentration results.
A205
  • Varies from peptide to peptide; approximate range 0.002–10.74 mg/mL 
  • Measures peptide backbone absorbance at 205 nm. For peptides that lack or have few Trp and Tyr residues.
  • Scopes method option can be used to measure proteins that have significant amounts of Trp and Tyr.
  • More sensitive than A280 since A205, molar absorptivity is high.
  • High salt protein buffers such as PBS and TE absorb in the low UV. Use a low salt buffer like the Brij® buffer diluted to 0.01%.
Pierce 660
  • 50–2000 μg/mL BSA (15:1 reagent: sample vol)
  • 25–1,000 μg/mL BSA (7.5:1 reagent: sample vol)
  • The most accurate, fast, and easy colorimetric assay. 
  • Compatible with most detergents, reducing agents and Laemmli loading buffer. 
  • Assay done at room temperature with short incubation times.
  • Reagents stored at room temperature. 
  • Requires standard curve. Expanded linear range for standard curve compared to Bradford.
  • Less protein-to-protein variation than the Bradford assay.
Bradford
  • 100–1,000 μg/mL BSA (30:1 reagent: sample vol)
  • 15–100 μg/mL BSA (1:1 reagent: sample vol)
  • Measures Coomassie Blue shift at 595 nm when bound to proteins. 
  • Fast and easy assay done at room temperature.
  • Requires standard curve.
  • Surfactants may cause reagent precipitation. 
  • Coomassie-dye binding assay exhibits twice as much protein-to-protein variability as the BCA assay.
BCA
  • 125-2,000 μg/mL BSA (20:1 reagent: sample vol) 
  • 20–200 μg/mL BSA (1:1 reagent: sample vol)
  • Measures Cu-BCA chelates at 562 nm formed in the presence of proteins. 
  • Often used for more dilute samples.
  • Compatible with most surfactants at concentrations up to 5%.
  • Less protein-to-protein variation compared to Coomassie methods.
  • Requires standard curve.
  • Reducing agents, copper chelators and high-capacity buffers can interfere with assay.
Lowry
  • 200-1,500 μg/mL BSA
  • Measures Cupric sulfate-tartrate complexes at 650 nm.
  • Modified Lowry at 750 nm can be run as a custom method
  • Complexes can be measured from 650-750 nm.
  • Requires standard curve.
  • Potassium ions and detergents precipitate reagent.
  • Chelating agents, reducing agents and free thiols interfere with this assay.
Protein methods by NanoDrop instrument
Method
NanoDrop Ultra/UltraC/Ultra FL/UltraC FL
NanoDrop Eight
NanoDrop Lite Plus
A280
A280 Pro No No
A205
No
Pierce 660
No
Bradford
No
BCA
No
Lowry
No
A260/A280
Acclaro Contaminant Identification
No
Customizable protein editor
No
Absorbance at 280nm provides fast protein and peptide quantification
Purified protein samples can be accurately measured using direct absorbance at 280 nm, which is mostly due to the aromatic chains on the Trp and Tyr amino acids. Protein A280, a preconfigured application in our software, is the most popular quantification method because it is fast and simple, requires no reagents or standard curves, and consumes very little sample.
Unlike nucleic acids, each pure protein has a unique Beer-Lambert extinction coefficient based on its amino acid sequence. For accurate results, select the correct sample type from the menu or enter its protein extinction coefficient manually. The NanoDrop Ultra and NanoDrop Eight Protein Editor feature allows you to save the extinction coefficients of specific proteins so that you can customize your Sample Type options.
In addition to concentration, NanoDrop spectrophotometers and fluorometers can provide information about contaminants in the sample. Acclaro Sample Intelligence Technology (included on the NanoDrop Ultra and NanoDrop Eight instruments) uses mathematical algorithms to detect nucleic acids in protein samples and correct the concentration result as needed. Sample purity can also be assessed by measuring its A260/A280 ratio; a value >1 may indicate nucleic acid contamination in the protein sample.
A205 is a direct measurement option, even for tyrosine and tryptophan-free biomolecules
What if your protein or peptide lacks Trp and Tyr residues and thus cannot be measured using the A280 application? Since a protein’s peptide backbone absorbs light at 190–220 nm, peptides that lack Tyr or Trp residues can be quantified using absorbance at 205 nm. Protein A205 is a preconfigured application in NanoDrop software.
Proteins that have significant amounts of Trp and Tyr can also be quantified using the A205 application by selecting the Scopes method option. In fact, the A205 method has some advantages over the A280 protein method such as lower protein-to-protein variability (because A205 extinction coefficients are not based on amino acid composition) and higher sensitivity (because of proteins’ high molar absorptivity at 205 nm). Although technical limitations made these measurements difficult in the past, NanoDrop sample-retention technology and low stray light performance have simplified quantification of small amounts of protein by A205 methods.
One limitation of the A205 method is that many commonly used protein buffers have absorbance at 205 nm. Before using this technique, we recommend checking the protein buffer for any contribution to the absorbance at 205 nm.
Colorimetric assays for proteins in cell extracts or lysates
Proteins in complex mixtures such as cell extracts or lysates are best measured using a protein colorimetric assay such as the Bradford, BCA, Lowry, or Pierce 660nm assay. These assays provide protein-specific concentrations, avoiding absorbance from cell components that absorb in the UV range and would inflate A280. These and other applications are preconfigured on selected NanoDrop instruments.
BCA protein assay on the NanoDrop Ultra Spectrophotometer and Fluorometers
BCA assay results show total protein concentration (3 green circles) and the standard curve.
Quantification of labeled proteins
Labeled antibodies or other fluorescently labeled proteins and metalloproteins can be quantified using the Proteins & Labels preconfigured software application that delivers the protein as well as the label concentration (up to 2 dyes). This application is available as a preconfigured application on NanoDrop Ultra Spectrophotometers and Fluorometers, and NanoDrop Eight Spectrophotometers.
Acclaro Pro – enhanced accuracy
Add-on Thermo Scientific™ Acclaro™ Pro software for when exceptional spectrophotometric accuracy is required. The most popular measurement applications are unlocked from the factory providing ± 5% Error throughout the absorbance dynamic range.
Key benefits include:
Accurate High Concentration Measurements – absorbance measurements within 5% error up to 550 Abs (10 mm equivalent) – up to 400 mg/mL IgG or 18,000 ng/µL ssDNA.
Quick Dilution-Free Process – take triplicate measurements on the pedestal in 2 minutes
Enhanced calibration – bundle purchases get an additional calibration from the factory to calibrate the instrument to provide exceptional accuracy within the Acclaro Pro applications.
Preprogrammed Acclaro Pro applications on the NanoDrop Ultra Spectrophotometers and Fluorometers
Tap to select the application you need from the Acclaro Pro home screen.
Quality control laboratories
Quality assurance and quality control are more important now than ever before. In busy QC labs, NanoDrop spectrophotometers and fluorometers can help make the analysis of sample concentration and purity reproducible, routine, and reliable. Integrating next-generation sample quality assessment into your workflow makes a difference because time is precious and the quality of all your samples is paramount.
mRNA vaccine quality control
The mRNA vaccine against COVID-19 was the first mRNA vaccine to get FDA approval. Its success has re-ignited interest in using mRNA to deliver vaccines against a myriad of diseases including cancers. In vaccine production, quality checks of sample concentration and purity are useful at several steps in the process.
  • In the sequencing stage, both the quality and quantity of the starting material are crucial for accurate results.
  • In plasmid production, the purified plasma must be checked for genomic DNA and other contaminants.
  • In in vitro transcription, the purified mRNA must be checked for purity before the mRNA solution is filled into the vaccine vials.
mRNA vaccine production workflow
The NanoDrop Ultra spectrophotometers and fluorometers, and NanoDrop Eight spectrophotometers can serve as quality control checkpoints at several stages in the production of mRNA vaccines.
View our on-demand webinar to see how the NanoDrop spectrophotometers can readily be deployed for all these tasks. And, with software available to aid compliance with FDA 21 CFR Part 11 regulations, these instruments are ready to be implemented into the mRNA vaccine workflow at a GMP laboratory.
Webinar: mRNA vaccines: development, manufacturing, and how NanoDrop Eight can help
This webinar briefly discusses mRNA vaccine history and workflows and examines how NanoDrop spectrophotometers can aid researchers and production engineers in assessing nucleic acid concentration and purity while complying with 21 CFR Part 11 regulations.
Compliance in pharmaceuticals, biotechnology, and other regulated environments
Our trusted, premier NanoDrop Ultra spectrophotometers and fluorometers, and NanoDrop Eight spectrophotometers can be used in regulated environments such as pharmaceuticals and biotechnology. These models can help you quantify and qualify DNA, RNA, and protein samples of only 1–2 µL in seconds and obtain full-spectrum data to confirm that samples are qualified for downstream applications.
In addition, optional Thermo Scientific SciVault or Thermo Scientific SciVault 2 software helps your lab stay compliant with US FDA data regulations every step of the way. These software modules enable regulatory compliance with part 11 of the US FDA’s Title 21 of the Code of Federal Regulations pertaining to electronic records and signatures (21 CFR Part 11).
The following sections show selected features of the SciVault 2 software for the NanoDrop Ultra Spectrophotometers and Fluorometers, which include user account management, user account access, audit trail capability, electronic signatures, automated data back-up, and more. SciVault software for the NanoDrop Eight instrument provides similar functionality.
How SciVault 2 software meets US FDA Title 21 CFR Part 11 requirements
Requirement: Limit system access to authorized individuals.
Feature: Administrators grant access through the SciVault 2 software. User account access is managed through the local instrument user management tool or through the Windows login system when running the software from a PC.
Requirement: Signed electronic records have printed name, date and time stamp, and meaning of signature.
Feature: Experiments are signed immediately after completion. Users can revise, review, and approve experiments from the History page.
Requirement: Secure, computer-generated, time-stamped audit trails to record operator actions and entries.
Feature: The Audit Logs feature creates an electronic audit trail of actions such as logons, data creation, signatures, and more. Audit logs are available as needed.
Pharmaceuticals compliance webinar
The ever-changing regulatory environment faced by manufacturers in pharmaceuticals and other regulated industries can cause confusion and doubts about compliance. Instrument systems designed to meet regulatory requirements can relieve the burden on manufacturers. How do they do it? Watch our webinar on maintaining compliance in pharma QA/QC labs to see how two different instrument platforms—the Thermo Scientific Nicolet Summit FTIR Spectrometer (for raw and finished material QA/QC) and the NanoDrop Spectrophotometer (for DNA, RNA, and protein quantification)— help labs assure compliance in their processes.
Webinar: Maintaining compliance in pharma QA/QC labs
Learn how new software tools are enabling drug companies to perform highly accurate QA testing, while ensuring data integrity throughout the drug development process.
Bacterial culture growth (OD600)
The growth of bacterial cultures can be monitored by measuring their optical density at 600 nm (OD600). Although this is a central technique in microbiology, these optical density measurements often contain very little true chemical absorbance. Instead, they are representative of the amount of light scattered away from the instrument’s detector by the bacterial suspension.
You can measure OD600 on the NanoDrop Ultra spectrophotometers and fluorometers, and NanoDrop Eight spectrophotometers using the OD600 preprogrammed application. The software uses the Beer-Lambert equation and a user-entered cell number conversion factor to automatically convert the OD600 value into the number of cells per milliliter.
Note that on the NanoDrop UltraC NanoDrop UltraC FL instrument, the pedestal and cuvette options will give different OD600 values. You can use a conversion factor to compare between pedestal and cuvette measurements.
Time-based kinetic measurements
On the NanoDrop UltraC and NanoDrop UltraC FL instrument, you can make time-based kinetic measurements on samples in cuvettes. You can designate up to three wavelengths from 190–850 nm for continuous absorbance monitoring at user-defined intervals in up to 5 stages. Cuvette measurements offer an extended lower detection limit and an optional 37°C heater and micro-stirrer.
You can create, edit, and save kinetics experiments as custom methods.
Sample kinetics results
NanoDrop Ultraᶜ and Ultraᶜ FL for absorbance at 250, 260, and 325 nm at user-defined intervals over 2 minutes.
Custom methods for NanoDrop spectrophotometers and fluorometers
NanoDrop spectrophotometers and fluorometers come with pre-loaded methods for nucleic acid quantification (like dsDNA, ssDNA, and RNA) and protein quantification (like A280, BCA, and Bradford) as described in the sections above. You can also create custom methods on the NanoDrop Ultra spectrophotometers and fluorometers or NanoDrop Eight spectrophotometer to perform UV-Vis or other user-defined measurements to suit your needs, allowing the instrument to function as a conventional spectrophotometer. Custom methods can monitor and report up to 40 wavelengths from 190–850 nm and can be set up directly from the touchscreen or PC control software. Custom methods provide the flexibility to use your instrument for additional applications.
  • Use predefined custom methods to analyze samples such as nanoparticles, chlorophyll, hemoglobin, glucose, and proteins via BCA assays.
  • Create new custom methods to analyze your special samples and save the methods for future use.
Measure forms of hemoglobin using a custom method
As a case example, a spectrophotometer may be used to quantitate various forms of hemoglobin using the appropriate extinction coefficient and peak absorbance value.
  • Oxyhemoglobin contains bound oxygen and exhibits absorbance peaks at 414 nm, 541 nm, and 576 nm.
  • Deoxyhemoglobin does not contain bound oxygen and exhibits an absorbance peak at 431 nm.
  • Methemoglobin cannot bind oxygen due to containing iron in the ferric state and exhibits an absorbance peak at 406 nm.
We created a custom method for the NanoDrop Ultra instruments to measure the absorbance of hemoglobin at 406, 414, 431, 541, and 576 nm—the five wavelengths needed to differentiate oxyhemoglobin, deoxyhemoglobin, and methemoglobin. 
Education: NanoDrop spectrophotometers and fluorometers in the classroom
Access to real-world instrumentation is key to preparing and inspiring the next generation of scientists. Hands-on experience helps make science more approachable and scientific principles more memorable.
In particular, UV-Vis spectroscopy is a valuable tool for studying macromolecules such as proteins and nucleic acids in the biology classroom. The simplicity and affordability of NanoDrop instruments makes them highly suitable for use in undergraduate and even secondary science curricula. Experience with NanoDrop spectrophotometers can support lessons in genomics, immunology, microbial cell growth, enzyme kinetics, and the structure and function of the fundamental components of life.
This webinar documents how two high school teachers have integrated NanoDrop One Spectrophotometers into their classes and workshops.
Webinar: NanoDrop innovations and the classroom: Inspiring the next generation of scientists
Access to the latest technology can make a world of difference in education today. Hear how schoolteachers are integrating NanoDrop instruments into their classrooms and labs, providing students with the opportunity to learn on industry-leading technology.
For Research Use Only. Not for use in diagnostic procedures.
Brij is a registered trademark of UNIQEMA AMERICAS LLC. All other trademarks are the property of Thermo Fisher Scientific Inc., and its subsidiaries.