Search
CyQUANT XTT and MTT Assays for Cell Viability |
XTT and MTT assays for cell viability are simple and optimized colorimetric-based assays that function to evaluate cell proliferation and viability on microplate readers. These assays use the cellular redox potential in metabolically active cells to convert a tetrazolium-based salt reagent—MTT or XTT—to a brightly colored formazan product.
| CyQUANT MTT Cell Viability Assay | CyQUANT XTT Cell Viability Assay | |
|---|---|---|
| Assay type | Colorimetric mix-and-read assays using the cellular redox potential in metabolically active cells | |
| Use |
|
|
| Components |
|
|
| Mechanism of detection | Conversion of water-soluble MTT to an insoluble purple-colored formazan | Conversion of water-soluble XTT to a water-soluble orange-colored formazan![]() |
| Solubility | Insoluble formazan; requires additional solubilization steps with SDS or DMSO | Soluble formazan; does not require additional solubilization steps |
| Sensitivity | ![]() | ![]() |
| Incubation time | 8+ hours | 4 hours |
| Continuous assay | No | Yes |
| Scalability | 96-well plate | 96- and 384-well plate |
| Reproducibility | Moderate | High |
| Absorbance (nm) | 570 nm | 450 nm and 660 nm |
| Cat. No. | V13154 | X12223 |
The following stand-alone product is also available: MTT 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (Cat. No. M6494).
See also: MTT Cell Viability Assay Kit Protocol
An MTT assay is a simple method for quantitatively determining cell number using standard absorbance microplate readers. It is a well-established assay that measures for cellular viability and proliferation.
The CyQUANT MTT cell viability assay uses the cellular redox potential of metabolically active cells to reduce water-soluble MTT to an insoluble purple formazan. The formazan is then solubilized with sodium dodecyl sulfate (SDS, included in the assay) and measured on a microplate reader at 570 nm (Figure 1). The concentrated formazan is directly correlated with the number of metabolically active cells (Figure 2).
Figure 2. Quantitation of Jurkat cells using the CyQUANT MTT Cell Viability Assay Kit. Cells in the parent culture were counted, diluted to the indicated densities in 100 µL volumes, then transferred to wells of a microplate, and incubated for four hours with CyQUANT MTT Cell Viability Assay to allow time for adsorption. Absorbance was measured at 570 nm using a microplate reader. Each data point represents the mean value of samples in triplicate. The inset shows the data plotted for the lower cell numbers.
To use the MTT assay, prepare the MTT stock solution and the SDS-HCl solution, add the MTT solution, incubate for four hours at 37ºC, add the SDS-HCl solution, incubate for an additional 4–18 hours in a humidified chamber, then read the absorbance at 570 nm. The use of DMSO as a solubilizing agent may be substituted for the SDS-HCl solution with incubation reduced to ten minutes at 37ºC and absorbance read at 540 nm (not 570 nm).
See also: MTT Cell Viability Assay Kit Protocol
CyQUANT XTT cell viability assay is a continuous assay for quantitatively determining cell viability and proliferation using the cellular redox potential in live cells. XTT has shown to be particularly useful for high throughput screening of antiviral and antitumor agents, and for assessing the effects of cytokines on cell proliferation.
The XTT assay is an easy-to-use, optimized assay that contains the XTT reagent and an Electron Coupling Reagent. The XTT assay uses the cellular redox potential of metabolically active cells to reduce water-soluble XTT to a water-soluble orange-colored formazan. The Electron Coupling Reagent, an electron mediator, captures readily available electrons on the plasma membrane to form a reactive intermediate that aids in the reduction of XTT (Figure 3).
The water-soluble formazan product allows for easier quantification of cellular activity and helps reduce overall assay time with a recommended four-hour incubation for optimal results; however, the assay can be completed in as little as two hours without affecting sensitivity. The Electron Coupling Reagent increases sensitivity and improves consistency, displaying an up to an eight-fold increase in signal-to-background ratios (Figure 4). Further, the CyQUANT XTT assay does not require cell lysis and uses noninvasive probes allowing for the continuous analysis of cell health with additional functional reagents.
See BioProbes article: An Absorbance-Based Assay for Cell Health and Proliferation
See poster: New and improved cellular health evaluation of 2D and 3D cellular models using microplate reader assays
Figure 4. Viability determination using multiple methods. A549 cells were plated at various concentrations (x-axis). After an overnight incubation, the manufacturer’s instructions were followed for either the CyQUANT XTT Cell Viability Assay, the Cell Proliferation Kit II (Sigma), or MTT Reagent. For the XTT-based assays, the absorbance was read at 450 and 660 nm using the Varioskan LUX instrument. Cell Viability using MTT Reagent required a 540 nm absorbance measurement. The specific absorbance signals were determined and signal-to-background (wells with no cells) ratios were generated.
Using the CyQUANT XTT cell viability assay is simple. Thaw the XTT Reagent and Electron Coupling Reagent, mix to prepare the working solution, add the solution to your cells, incubate for four hours at 37ºC, and read the absorbance.
It is important to use the XTT/Electron Coupling Reagent immediately after preparation as performance and sensitivity will be reduced if the solution is kept at room temperature for an extended period of time or subject to freeze/thaw cycles. To ensure assay performance, the assay consists of separate XTT and Electron Coupling Reagents that should be kept at –20ºC until use.
The XTT-specific absorbance is measured at 450 nm. The 660 nm absorbance reading is used to eliminate the background signal contributed by cell debris or other non-specific absorbance.
CyQUANT XTT and MTT assays were used to generate drug-dose response curves in cells exposed to gambogic acid, a xanthanoid compound known for its research into anticancer effects, including inhibiting cell growth by inducing apoptosis and autophagy [1]. Comparative drug dose response curves for research showed IC50 values were similar between the XTT and MTT assay (Figure 5).
Figure 5. Comparison of CyQUANT XTT and MTT assay sensitivity. The CyQUANT XTT Cell Viability Assay and a commercially available MTT assay were used according to their respective manufacturers’ protocols to generate drug-dose response curves for A549 cells exposed to increasing concentrations of gambogic acid for ~18 hours at 37°C and 5% CO2. The IC50 value for gambogic acid generated with the CyQUANT XTT assay (3.28 μm) was similar to that obtained with the MTT assay (2.76 μm). Absorbance of the XTT assay product is reported as the absorbance of the sample at 450 nm minus the absorbance of the sample at 660 nm minus the absorbance of the blank at 450 nm. Absorbance of the MTT assay product is reported as the absorbance of the sample at 540 nm minus the absorbance of the blank at 540 nm. All measurements were made using a Thermo Scientific Varioskan LUX Multimode Microplate Reader, and data were analyzed using GraphPad Prism software.
High-sensitivity fluorescence detection for 6-1,536 samples can be quickly performed on the Varioskan ALF or Varioskan LUX Multimode Microplate Reader using Invitrogen reagents to enable optimal detection. Take advantage of automatic dynamic range selection to get optimal gain settings for each individual well and automation capabilities for even higher throughput.
Cell Analysis Learning Center—Find educational resources such as application notes, webinars, videos, articles, and more that cover the use of many of our reagents and kits for cell analysis.
Fluorescence SpectraViewer—Online tool for visualization of the excitation and emission of fluorescent reagents; allows for checking spectral compatibility for multiple fluorophores.
Cell Analysis Support Center—Find technical information, tips and tricks, and answers to everyday problems.
For Research Use Only. Not for use in diagnostic procedures.