The MEGAscript® Kit is the most frequently cited large-scale transcription kit, and with good reason. It enables the consistent transcription of high yields of full-length RNA (Figure 1). Ambion offers a complete line of products specifically optimized to generate the following products:
  • capped RNA
  • dsRNA transcripts
  • uncapped RNA
  • amplified RNA
  • short RNA transcripts
  • milligram quantities of transcript
Ordering Information
Catalog # Name Size Price (USD) Qty
AM1334 MEGAscript™ T7 Transcription Kit, 40 Reactions Each
406.78
Special offer
Online exclusive
Ends: 12-Sep-2025
473.00
Save 66.22 (14%)
AM1345 mMESSAGE mMACHINE™ T7 ULTRA Transcription Kit Each
467.68
Special offer
Online exclusive
Ends: 12-Sep-2025
632.00
Save 164.32 (26%)
AM1333 MEGAscript™ T7 Transcription Kit, 25 Reactions Each
277.68
Special offer
Online exclusive
Ends: 12-Sep-2025
312.00
Save 34.32 (11%)
AM1344 mMESSAGE mMACHINE™ T7 Transcription Kit Each
567.60
Special offer
Online exclusive
Ends: 12-Sep-2025
660.00
Save 92.40 (14%)
AM1751 MessageAmp™ II aRNA Amplification Kit, 20 Reactions Each
2,420.00
AM1354 MEGAshortscript™ T7 Transcription Kit Each
419.19
Special offer
Online exclusive
Ends: 12-Sep-2025
471.00
Save 51.81 (11%)
AM1330 MEGAscript™ SP6 Transcription Kit, 40 Reactions Each
418.60
Special offer
Online exclusive
Ends: 12-Sep-2025
460.00
Save 41.40 (9%)
AM1340 mMESSAGE mMACHINE™ SP6 Transcription Kit Each
633.36
Special offer
Online exclusive
Ends: 12-Sep-2025
696.00
Save 62.64 (9%)
AM1626 MEGAscript™ RNAi Kit Each
551.04
Special offer
Online exclusive
Ends: 12-Sep-2025
672.00
Save 120.96 (18%)
AMB13345 MEGAscript™ T7 Transcription Kit, 200 reactions Each
1,315.28
Special offer
Online exclusive
Ends: 12-Sep-2025
1,604.00
Save 288.72 (18%)
AM1338 MEGAscript™ T3 Transcription Kit Each
440.55
Special offer
Online exclusive
Ends: 12-Sep-2025
495.00
Save 54.45 (11%)
AM1348 mMESSAGE mMACHINE™ T3 Transcription Kit, 25 Reactions Each
581.36
Special offer
Online exclusive
Ends: 12-Sep-2025
676.00
Save 94.64 (14%)
MEGAscript: Consistent, High Yields of RNA Transcripts
Using the MEGAscript Kit, you can produce over 120 µg of RNA from 1 µg of DNA template. This is 10-50 times the amount of RNA produced by conventional transcription (Figure 2). Reactions are versatile and can be manipulated to include specialized reagents such as modified nucleotides or additional polymerase. In addition, Ambion has optimized the transcription of short (20-500 nt) DNA templates with the MEGAshortscript™ Kit. This patented technology was designed to maximize the yield of short RNA transcripts from plasmid, oligonucleotides, and PCR product templates.
Figure 1. Electropherogram of RNA Transcripts of Different Sizes. In vitro transcription was performed using 1, 4, or 9 kb templates with Ambion's MEGAscript® Kit according to protocol. 20 µl reactions were diluted to 100 µl and 1 µl was assessed on the Agilent 2100 bioanalyzer. The single primary peak is indicative of product quality while the area under the peak is related to yield.
Figure 2. MEGAscript® vs. Conventional In Vitro Transcription Reactions. 20 µl transcription reactions containing 1 µg template, T7 MEGAscript components and [ 32P]UTP, were incubated at 37°C. 1 µl aliquots were removed at 1 hr intervals and acid precipitable cpm determined by scintillation counting.
mMESSAGE mMACHINE®: Translatable Capped RNA
For high yields of capped RNA, MEGAscript technology is incorporated into the mMESSAGE mMACHINE® Kit. This kit contains an optimized ratio of cap analog to GTP, for the best trade off between yield and proportion of capped transcripts (80% capped). Furthermore, you can obtain 100% translatable capped RNA with the mMESSAGE mMACHINE T7 ULTRA Kit. This kit uses a unique cap analog that only incorporates in the functional, translatable orientation, resulting in the synthesis of capped RNAs that are 100% translatable. 
MEGAscript® RNAi: dsRNA Synthesis for RNAi
In one complete kit, you can generate ready-to-use double-stranded RNA (dsRNA) for RNAi experiments in non-mammalian systems. Unlike competing products, the MEGAscript RNAi Kit provides reagents to eliminate DNA template, single stranded RNA, and free nucleotides. The MEGAscript RNAi Kit allows researchers to synthesize 50 to 80 µg or more of dsRNA, up to 1.2 kb in length, in a single transcription reaction. 
MessageAmp™ II: RNA Amplification
Amplify RNA with MEGAscript technology using the MessageAmp™ II aRNA Amplification Kit. The MessageAmp II Kit can amplify as little as 100 ng of input RNA in a single round for GeneChip® analysis. A rationally engineered M-MLV RT, ArrayScript™, is provided in the kit to maximize yields of full-length cDNA. cRNA synthesis is powered by MEGAscript technology. These features result in high labeling efficiency, yielding consistently high percentages of Present Calls, and low 3'/5' ratios in GeneChip analysis. This procedure is compatible with biotin, cyanine dye (Perkin Elmer), and Cy™Dye (Amersham Biosciences) labeling either during or after aRNA amplification. For additional information, see RNA Amplification Just Got Better.
Bulk Transcription - Milligram Quantities of RNA
If you have a project or application that requires large amounts of RNA transcripts, Ambion can meet your needs with its bulk and custom products. Our MEGAscript technology has been optimized to produce milligram quantities of RNA.