Search
Maintain your Thermo Scientific NanoDrop instruments, streamline your research, and increase your productivity with these helpful tools and accessories.
NanoDrop Ultra ds DNA Broad Range (BR) Fluorescence Assay Body
Enables broad range dsDNA sample quantification via fluorescence. Concentrated assay reagents requiring dilution. Origional sample concentration range: 10 to 1,000 ng/ul. Up to 250 Reactions.
NanoDrop Ultra RNA Broad Range (BR) Fluorescence Assay Body
Enables broad range RNA sample quantification via fluorescence. Concentrated assay reagents requiring dilution. Original sample concentration range: 10 to 1,000 ng/uL. Up to 250 Reactions
PV-1/PV-8 Performance Verification Solution/Kit
While most users will never need to have their NanoDrop instruments calibrated, we recommend verifying with this solution every 6 months to help ensure measurement accuracy and reliability. For use with NanoDrop Ultra/UltraC, Ultra FL/UltraC FL, NanoDrop One/OneC, NanoDrop Eight, and NanoDrop Lite Plus instruments.
PR-1 Reconditioning Kit
Compound and swabs to recondition NanoDrop sample measurement pedestals to a hydrophobic state.
FL-1 Fluorescence Verification Kit
Reagent-based assay that tests the performance of the NanoDrop Ultra FL and NanoDrop UltraC FL. Includes: Blank Reagent, Diluent, Green Fluorescence Reagent - designed to test the blue excitation LED and the green fluorescence detection, Far-red Fluorescence Reagent - designed to test the red excitation LED and the far-red fluorescence detection.
CF-1/CF-8 Calibration Check Fluid/Kit
Concentrated, aqueous potassium dichromate (K2Cr2O7) solution for confirming calibration. For use with NanoDrop 1000, NanoDrop 2000/2000c, NanoDrop Lite, and NanoDrop 8000 instruments.
Printer Accessory
An optional docking printer for your NanoDrop Lite Plus or NanoDrop Lite instrument allows you to print data on freezer-compatible labels.
USB WiFi and Bluetooth Dongle
Enables wireless network connection and Bluetooth to the NanoDrop Ultra on-board instrument software.
Instrument Dust Cover
Transparent plastic dust cover protects the NanoDrop Ultra models against dust and any liquid exposure while not in use.
Starna Scientific™ DNACON10X-SC2 Reference Solution
A A260/A280 reference solution aimed at customers wishing to confirm the performance of their NanoDrop™ microvolume UV-Vis spectrophotometer.
NanoDrop spectrophotometer accessories and consumables by instrument
| Product | ND Ultra | ND UltraC | ND Ultra FL | ND UltraC FL | ND Eight | ND Lite Plus | ND One | ND OneC | ND 3300 | ND 2000 /2000c |
ND 8000 | ND Lite | Cat. No. |
PV-1 Performance Verification Solution |
✓ | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | CHEM-PV-1 | |||||
| PV-8 Performance Verification Kit | ✓ | CHEM-PV-8 | |||||||||||
| PR-1 Reconditioning Kit | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | CHEM-PR1-KIT |
| FL-1 Fluorescence Verification Kit | ✓ | ✓ | CHEM-FL1-KIT | ||||||||||
| ND Ultra dsDNA BR Fluorescence Assay | ✓ | ✓ | NDU-FL-DSDNA-BR | ||||||||||
| ND Ultra RNA BR Fluorescence Assay | ✓ | ✓ | NDU-FL-RNA-BR | ||||||||||
| CF-1 Calibration Check Fluid | ✓ | ✓ | CHEM-CF-1 | ||||||||||
| CF-8 Calibration Check Kit | ✓ | CHEM-CF-8 | |||||||||||
| Cuvette Qualification Filter Kit | ✓ | ✓ | ✓ | ✓ | 840-257300 | ||||||||
| Starna Sci DNACON10X-SC2 | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ | 869-193700 | |
| Printer Accessory | ✓ | NDLP-PRNTR-MOD |
|||||||||||
| ✓ | 840-218900 | ||||||||||||
| Printer Paper | ✓ | ✓ | 714-0174700 | ||||||||||
| USB Wi-Fi & Bluetooth Dongle | ✓ | ✓ | ✓ | ✓ | NDU-WIFI-STICK | ||||||||
| Dust Cover | ✓ | ✓ | ✓ | ✓ | NDU-INST-COVER |
Extract and purify DNA, RNA, proteins, and cells with Thermo Scientific™ KingFisher™ instruments. Reduce overall processing time and errors associated with manual sample preparation while increasing the yield and reproducibility of your results. Access easy-to-follow protocols and maintain consistent results across runs and different users for sensitive downstream applications.