Search
Researchers are navigating the modern complexities of genetics in healthcare. Thermo Fisher helps make drug response research more precise with preemptive pharmacogenomic arrays. Modern medication is responsible for saving millions of lives a year; yet a variety of factors such as lifestyle, environment, and genes can cause a medication that is beneficial for one individual to be harmful for another. An adverse drug reaction (ADR) can occur when a prescribed drug is incompatible with the patient.
Pharmacogenomics (PGx) is an exciting and powerful emerging field that can help improve health care outcomes. Pharmacogenomics provides meaningful data that can help prevent adverse drug reactions, deliver beneficial treatments faster, and lower medical costs.
Pharmacogenomics research has often required the use of more than one molecular assay, increasing the cost and time to result. The PharmacoScan and PharmacoFocus assays help overcome this concern by interrogating SNPs, insertions/deletions (indels), and CNVs in a single-assay workflow. Key markers in pseudogenes like CYP2D6 are amplified by multiplex PCR (mPCR) in the assay.
The Axiom PharmacoScan Solution offers broad coverage of industry-relevant, multi-ethnic content and the capability to address markers in complex genes using an array-plate platform that has superior lot-to-lot reproducibility. The PharmacoScan Solution targets functional variations in the key genes involved in the absorption, distribution, metabolism, and excretion (ADME) of commonly prescribed medications and can help reduce the lengthy timelines and complexities within the drug discovery process.
Unlike solutions that analyze only high-evidence markers, the Axiom PharmacoScan Solution presents a more comprehensive view for genotyping for pharmacogenomic risk by analyzing over 4500 high- to low-evidence markers within nearly 800 genes in a single assay. The solution includes CNV calling and interrogating predictive markers in complex genes.
The content of the PharmacoScan array (Figure 1) includes:
Fig 1. Distribution of consortia and sources used to select the ADME markers for the PharmacoScan Solution.
The Axiom PharmacoFocus Solution is ideal for preemptive pharmacogenomics with the flexibility of a multi-gene test panel aligned with your pharmacogenomics research, whether it takes place in labs, academic hospitals, healthcare centers, or the pharma industry.
The Axiom PharmacoFocus assay provides comprehensive coverage of high evidence content (88% of variants in PharmGKB highest levels of evidence) with 2,000 markers in 150 genes covering population diversity. This unique solution offers total flexibility to choose few or all from a comprehensive set of 2,000 high-evidence ADME variants to generate high accuracy pharmacogenomic SNP, CNV, and HLA typing, all in a single workflow at lower cost. The array has been designed to enable accurate genotyping of even technically challenging ADME variants in complex genes through a gene-specific amplification step.
With over 35% of high-evidence functional variants implicated in toxicity and adverse drug reactions (ADRs) the Axiom PharmacoFocus Array is ideal for research on population-scale preemptive pharmacogenomic strategies or early-phase clinical research to support better research outcomes.
| Category | Number of Variants |
| Pharmacogenomics (PGx) variants | >2100 |
| Number pf PGx genes covered | 150 |
| PGx variants by PharmGKB levels of evidence | |
| 1 | 114 |
| 2 | 155 |
| 3 | 300 |
| 4 | 198 |
| Highly predictive markers in regions of high homology within CYP2C19, CYP2C9, CYP2D6, CYP1A2, CYP2A6, CYP2B6, GSTM1, and SULT1A1 | >110 |
| Variants for calling over 970 unique haplotypes across 66 gene regions | >850 |
| HLA—variants enabling HLA typing for 11 major histocompatibility complex loci (3 MHC Class I and 8 Class II) | >1,560 |
| TaqMan OpenArray catalog panel variants | >119 |
| Dutch Working Group Pharmacogenomic passport markers (2019) | 65 |
| Medicaid (CMS) reimbursement policy, including psychiatry panel | >19 variants in CYP genes and HLA types |
The Axiom PangenomiX Array is the most comprehensive array within the Axiom microarray catalog for studying disease susceptibility and risk, pharmacogenomics (PGx), and genetic factors underlying wellness and lifestyle in diverse populations. It offers >850,000 markers selected for high genomic coverage from the 1000Genomes Project Phase III and other broadly referenced public databases, including ClinVar, NHGRI-GWAS catalog, CPIC, and PharmaGKB. Publicly available imputation algorithms enable imputation to the whole genome.
The pharmacogenomics module includes:
The Axiom PangenomiX Array used in conjunction with the Axiom 2.0 Plus Assay can unlock over 80 critical star alleles associated with highly predictive markers in genes including CYP1A2,CYP2D6, CYP2B6, CYP2A6, SULT1A1, CYP2C19, and CYP2C8. This unique assay opens up the ability to genotype these important pharmacogenomic markers that are in highly homologous regions of the genome.
Axiom myDesign arrays allow flexible access to custom genotyping for small and large studies, supported by a world-class design team for fast design and delivery. With Axiom myDesign Genotyping arrays, you can create fully or semi-customizable genotyping arrays for agrigenomics and human genotyping applications. Based on the universal workflow of the Axiom Genotyping Solution, Axiom myDesign arrays offer unprecedented scale and flexibility to design an optimal array for your species of interest. Fully or semi-customized arrays containing 1,500 to 2.6 million markers can be designed and available in as few as six weeks after SNP lockdown. For more information, visit the Custom Arrays Page.
Axiom PharmacoScan Solution |
Axiom PharmacoFocus Solution |
Axiom PangenomiX Array |
| Throughput | ||
Available in 24 format or 96 format Includes the arrays, reagents, processing consumables, and analysis software needed for processing 88 samples on four 24-format array plates or 94 samples on each 96 format array plate
Each 24-format array plate can process 22 samples and 2 controls, and each 96 format array plate can process 94 samples and 2 controls |
Available in mini 96 Format Includes the array, reagents, processing consumables, and analysis software needed for processing 94 samples on each mini 96-format array plate.
Each mini 96 format array plate can process 94 samples and 2 controls |
Available in 96 Format Includes the array, reagents, processing consumables, and analysis software needed for processing 94 samples on each 96-format array plate.
Each 96 format array plate can process 94 samples and 2 controls |
| Pharmacogenomic markers | ||
| 4.5K | 2k | 5k |
| High evidence coverage | ||
225 markers (PharmGKB level 1A-2B markers as of May 2021) |
242 markers (PharmGKB level 1A-2B markers as of May 2021) |
233 markers (PharmGKB level 1A-2B markers as of May 2021) |
| CNV detection | ||
| 3 gene regions of CYP2A6, 3 gene regions of CYP2D6, GSTM1, GSTT1 (CN=0-3); UGT2B17 (CN=0-2); SULT1A1 (CN=0-4+) | 3 gene regions of CYP2A6, 3 gene regions of CYP2D6, GSTM1, GSTT1 (CN=0-3+); UGT2B17 (CN=0- 2+); SULT1A1 (CN=0-4+) | 3 gene regions of CYP2A6, 3 gene regions of CYP2D6, GSTM1, GSTT1 (CN=0-3+); UGT2B17 (CN=0-2+); SULT1A1 (CN=0-4+) |
| HLA typing | ||
| HLA types for 11 MHC loci’s including 4 HLA PGx relevant variants (HLA-A*31:01, HLA-B*15:02, HLA-B*57:01, HLA-B*58:01) | HLA types for 11 MHC loci's including 4 HLA PGx relevant variants (HLA- A*31:01, HLA- B*15:02, HLA- B*57:01, HLA B*58:01) | HLA types for 11 MHC loci's including 4 HLA PGx relevant variants (HLA-A*31:01, HLAB* 15:02, HLA-B*57:01, HLA-B*58:01) |
| Reporting capability | ||
| 946 star alleles/haplotypes in 66 gene regions | 970 star alleles/haplotype calls in 66 gene regions | 883 Star Alleles/haplotype calls in 63 gene regions |