Search
The most advanced and versatile multiplexing platform offers a compact footprint and compatibility with all Invitrogen ProcartaPlex multiplex immunoassays and Invitrogen QuantiGene Plex multiplex gene expression assays.
Available in two configurations, the Luminex xMAP INTELLIFLEX systems have 5.5 logs of dynamic range and an embedded PC with touchscreen interface. With 96- and 384-well plate formats available, you can process up to 153,600 tests/hour. The Luminex xMAP INTELLIFLEX DR-SE System features dual reporters (405 and 532 nm lasers), which are useful for a variety of applications including antibody neutralization studies and free versus bound drug assays. The Luminex xMAP INTELLIFLEX DR-SE System also features a side eject door which is compatible with automation.
Features |
xMAP INTELLIFLEX System | xMAP INTELLIFLEX DR-SE System |
5.5 logs of dynamic range—save time diluting samples and run more samples simultaneously with a broader dynamic range. |
✓ | ✓ |
Integrated touchscreen PC—convenient, compact and intuitive set-up on the built-in touchscreen. |
✓ | ✓ |
Built in barcode reader—faster set-up and the ability to better track large experiments. |
✓ | ✓ |
Automated start up, shutdown, and maintenance routines—increase your lab productivity with walk away routines. |
✓ | ✓ |
Dual-reporting capabilities—dual reporter system with a second violet (405 nm) laser and 4.5 logs of dynamic range to collect two parameters per analyte. |
✓ | |
Side eject door—compatible with automation to maximize throughput. |
✓ |
|
|
Optics |
|
Lasers, APDs, PMTs |
|
| Hardware | |
Flow cytometry–based lasers, APDs, PMTs |
|
| Footprint including PC (linear bench space) | |
58.4 cm x 61 cm x 76 cm |
|
| Weight (analyzer) | |
54.5 kg (120 lb) |
|
| Multiplex capacity | |
Up to 500 |
|
| Maximum available commercial assay plex | |
80 |
|
| Application | |
Protein/nucleic acid |
|
| Sample volume | |
6.3–80 µL (assay dependent) |
|
| Plate format | |
96-well or 384-well |
|
| Read time | |
96-well in ~20 min 384-well in ~75 min |
|
| Sensitivity (assay dependent) | |
0.06–1 pg/mL (immunoassays) |
|
| Reporter laser/optics | |
532 nm (green) and 405 nm (violet)* |
532 nm (green) |
| Dynamic range | |
≥5.5 logs (RP1) ≥4.5 logs (RP2) |
≥5.5 logs (RP1) |
| Operational validation | |
Calibration, verification, and fluidics |
|
| 21 CFR compliance | |
Yes |
|
| Automation | |
Yes |
|
| Analytics (software) | |
Data analysis apps for Luminex assays on the Thermo Fisher Connect Platform |
|
*The second reporter channel is only available in the xMAP INTELLIFLEX DR-SE model.
For Research Use Only. Not for use in diagnostic procedures.