Quality components designed for your liquid dispensing cassettes
The cassettes used with the Multidrop family of dispensers are robust, high-quality components designed for accurate and contamination-free liquid dispensing. They feature easy installation, compatibility with a wide range of reagents, and are available in autoclavable or disposable options, ensuring flexibility and reliability for diverse laboratory workflows.
Multidrop Combi Cassettes
Cassettes are essential components used in Thermo Scientific Multidrop reagent dispensers, designed to provide efficient and precise liquid dispensing in laboratory settings. These cassettes are available in various configurations to meet the needs of different applications, ensuring accurate volume delivery and compatibility with a wide range of reagents.
Standard Tube Dispensing Cassettes
Suitable for dispensing larger volumes (5 µL–2.5 mL) and are equipped with wide tubing for high-speed dispensing. They are designed with plastic covers, 40 cm silicone tubing, and polypropylene tip combs with 0.5 mm diameter tips. Extended versions with 200 cm tubing are available for automated systems.
Small Tube Dispensing Cassettes
Designed for smaller volume dispensing (0.5–50 µL) and come with either plastic or metal tips to accommodate different reagents. The cassettes feature 40 cm silicone tubing with 0.22 mm orifices and are available in standard and long tubing versions (200 cm).
SMART+ Dispensing Cassettes
Designed for use with the Multidrop Combi SMART+ dispensers, these cassettes include advanced features like radio frequency identification (RFID) tags that store lifetime dispensing data. They are compatible with a wide volume range (0.5 μL–2.5 mL) and come with either plastic or metal tips, making them suitable for a variety of reagent dispensing needs.
Multidrop Combi Cassettes Ordering Information
카탈로그 번호 제품명 제품 사이즈 Price 수량
24072670 Each
1,178,000
24072671 Each
4,954,000
24072672 Each
7,156,000
24072677 Each
1,665,000
24073290 Each
1,178,000
24073291 Each
5,164,000
24073295 Each
1,178,000
24073296 Each
4,880,000
24073293 Each -
24073298 Each -
N22700 Each
2,142,000
N22702 Each
2,218,000
N22706 Each
2,050,000
24070290 Each
725,000
24070297 Each
1,069,000
N22704 Each
2,235,000
Multidrop Pico 1 and 8 Cartridges
Cartridges are essential components used in Thermo Scientific Multidrop Pico dispensers, designed to provide efficient and precise liquid dispensing in laboratory settings. These cartridges are available in various configurations to meet the needs of different applications, ensuring accurate volume delivery and compatibility with a wide range of reagents.
1 channel
Pico 1
cartridge
4 channel 
Pico 8
cartridge
8 channel
Pico 8
cartridge
4 channel
Pico 200µl
8 channel
Pico 20µl
Multidrop Combi Cassettes Ordering Information
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.