Search
T Cell Expansion in Clinical Research |
Rely on the same proven technology platform throughout mouse and human stages of your T cell research. Obtain a 100- to 1000-fold expansion of T cells within 9–14 days, with no need for feeder cells or antigen presenting cells (APCs). T cells exhibit properties comparable to in vivoactivated T cells.
Dynabeads CD3/CD28 CTS (formerly known as Xcyte Dynabeads and Dynabeads ClinExVivo CD3/CD28) was developed to optimize ex vivo T cell expansion in translational research while preserving T cell viability and optimal immune function.
Designed to mimic in vivo activation and expansion, these ready-to-use Dynabeads are coated with antibodies directed against the TCR/CD3 and co-stimulatory CD28 receptors that are required for optimal T cell expansion.
The DynaMag CTS magnet is developed for optimal performance with the Dynabeads CD3/CD28 CTS technology, and allows you to work with large sample sizes (up to 10 L).
Figure 1. Diagram compares T cell activation with in vivo activation with Dynabeads.
In USA, Dynabeads CD3/CD28 CTS are available for clinical use only under an approved IND application. A Device Master File is held at the US Food and Drug Administration for cross-referencing in Investigational New Drug (IND) applications.
Dynabeads allow for specific capture of biomolecules while undesired molecules don't bind. Gentle magnetic separation leaves unbound molecules in the solution and bead-bound molecules ready for downstream analysis.
See Dynabeads Cell Isolation & Expansion support within the Cell Analysis Support Center for tips, troubleshooting help, and resources.
For Research Use Only. Not for use in diagnostic procedures.