Search
Amplification with a high-fidelity proofreading polymerase, such as Invitrogen™ Platinum™ SuperFi™ DNA Polymerase, leaves PCR products with blunt ends rather than 3′-A overhangs, due to the extensive 3′ to 5′ exonuclease activity. Therefore, the vectors supplied in the Invitrogen™ Zero Blunt™ TOPO™ PCR cloning kits contain blunt ends to offer more effective ligation. Choose from over 30 subcloning, sequencing, or long-fragment kits for 95% cloning efficiency in just 5 minutes.
Find tips, troubleshooting help, and resources for your cloning applications
Learn about PCR cloning strategies and subcloning approaches
Review the principles of PCR, components, cycling parameters, methods, and applications
For Research Use Only. Not for use in diagnostic procedures.