Search
Our newest multimode microplate reader is a versatile, entry level multimode microplate reader for most common absorbance, turbidimetric, fluorescence, and luminescence microplate assays. Photometric optical design is based on a xenon flash lamp and monochromator for enabling continuous wavelength selection from the UV through the visible range up to 1,000 nm. The broad wavelength ranges enable many common assays including nucleic acid and protein quantification, bacterial growth curves, ELISA, and cell viability.
In Varioskan ALF, fluorescence and luminescence measurements are performed with filter-based optics for optimal specificity and sensitivity. The system comes with three pre-installed excitation and emission filter sets suitable for many common fluorometric applications like Thermo Scientific Quant-IT DNA quantitation assays, PrestoBlue and AlamarBlue cell viability assays. Additionally, five open filter positions are available for customized fluorometric and luminometric measurements. Filter slots are easy to access from the instrument side and no additional tools are required for filter exchange. Also commonly used FRET and BRET assays are easy to set up and perform thanks to Varioskan ALF unique filter-based optics design. Explore technical specifications for factory-installed and accessory filter pairs for the Varioskan ALF Multimode Microplate Reader.
The automatic gain adjustment feature selects the ideal fluorometric or luminometric reading range for your instrument based on signal intensity in the well, eliminating the need to manually adjust measurement parameters. The results are consistent no matter what signals are measured.
In addition to being fast, flexible, and easy to use, the Varioskan ALF multimode reader also features:
| Category | Attribute | Description |
| Absorbance | Light source | Xenon flash lamp |
|---|---|---|
| Wavelength selection | Monochromator | |
| Wavelength range | 200–1,000 nm | |
| Bandwidth | 2.5 nm | |
| Wavelength setting resolution | 1 nm | |
| Wavelength accuracy | ±2 nm | |
| Stray light, @230 nm | <0.05% | |
| Linearity (absorbance), @450 nm | 96-well plate: 0–2.5 Abs, 2.0% | |
| Accuracy (absorbance), @450 nm | 96-well plate: For 0–2.0 Abs: ±(1.0% + 0.003 Abs) For 2.0–2.5 Abs: ±2.0% |
|
| Precision (absorbance), @450 nm | SD <0.003 Abs or CV <1.0%, whichever is greater | |
| DNA sensitivity @260 nm* | 96-well half area plate: <1 ng/μL | |
| Turbidimetric sensitivity* | 96-well plate: <5 formazin turbidity units (FTUs) | |
| Measurement speed** | 96-well plates Fast mode: ≤10 seconds Precision mode: ≤26 seconds |
|
| Spectral scanning speed | Fast mode: ≤16 seconds from 200–1,000 nm with 1 nm steps | |
| Precision mode: ≤200 seconds from 200–1,000 nm with 1 nm steps | ||
| Fluorescence intensity | Dynamic range | ≥6 decades |
| Sensitivity* | Black 384-well plate: 0.35 fmol/well FITC |
|
| Wavelength | Excitation wavelength: 200–710 nm Emission wavelength: 210–720 nm |
|
| Wavelength selection | Filter wheels with three factory filter pairs and five optional filter positions | |
| Factory installed filter pairs [wavelength (nm)/FWHM†(nm)] | Coumarin family filter pair | |
| Excitation 345/25 | ||
| Em 450/40 | ||
| FITC family filter pair | ||
| Excitation 485/15 | ||
| Emission 525/15 | ||
| Resorufin family filter pair | ||
| Excitation 555/15 | ||
| Emission 615/45 | ||
| Dual label measurements speed | <1 second | |
| Measurement speed, fixed dynamic range** | 96-well plates: ≤20 seconds | |
| Measurement time | 10–1,000 ms | |
| Luminescense | Wavelength selection | Filter wheel with no filter and five optional filter positions |
| Wavelength range | 200–720 nm | |
| Dynamic range | ˃6 decades | |
| Sensitivity* | White 96-well plate: <275 amol/well ATP | |
| Crosstalk (96-well white plate) | White 96-well plate: ≤0.1% | |
| Measurement speed, fixed dynamic range** | 96-well plates: ≤20 seconds | |
| Measurement time | 10 ms to 10 seconds | |
| Incubator | Temperature range | From ambient 2°C to 45°C (room temperature 25°C) |
| Liquid warm-up time | 96-well plate, 200 μL water/well: <60 minutes from 25°C to 37°C | |
| Mean temperature of wells at 37°C | ±0.5°C | |
| Plate temperature uniformity at 37°C | <1.0°C | |
| Shaker | Shaking modes | Linear, orbital, dual orbital |
| Shaking speeds | Low, medium, high | |
| Shaking type | Continuous or pulsed | |
| General features | Measurements modes | Endpoint, kinetic, spectra, kinetic spectra |
| Interface | PC software (SkanIt Software) | |
| Dimensions (D x W x H) | 61 x 34 x 40 cm (24 x 14 x 16 in) | |
| Weight | 25 kg (55 lbs) |
* Sensitivity specification values that represent the median of observed factory-tested values.
** Minimum kinetic interval time from A1 back to A1.
† FWHM: full width at half maximum.
Catalog number: VA000010C
For Research Use Only. Not for use in diagnostic procedures.