Search
Is your western workflow in need of a hero? Enlist these power pairs to come to the rescue.
Have the need for speed? Bolt Bis-Tris Plus gels offer the benefits of Bis-Tris chemistry, allowing you to load up to 60 uL of sample with run times of just 20 minutes. Tag-team with the iBlot 2 Dry Blotting System, which utilizes unique, ready-to-use dry transfer stacks for transfer times of only 7 minutes. Separate and transfer your proteins in a flash!
See Bolt Bis-Tris Plus gels
See the advantages of the iBlot 2 Dry Blotting System
With our innovative automated western blot processing system—the iBind Western Device—you can simply load primary antibody, secondary antibody, and wash solutions, then walk away. In less than 3 hours, your blot is ready for final detection. Solutions are processed using sequential lateral flow technology, with no batteries, shakers, trays, or timers required. Team up with our most versatile chemiluminescent HRP substrate with a great balance of sensitivity, signal duration, and signal intensity for a smooth and efficient immunodetection workflow.
Find chemiluminescent substrates
See what customers are saying about iBind Western Device
Find the perfect pair of primary and secondary antibodies for your western blot detection. We offer thousands of options in over 50 research areas such as cancer, epigenetics, immunology, and neuroscience. Find your match today.
You can trust the neutral pH of the NuPAGE Bis-Tris gels to resolve and maintain the integrity of your protein and provide you with straight lanes and publication-quality data. Combine them with the popular PageRuler Plus Prestained Protein Ladder (molecular weight range of 10–260 kDa) to monitor your gel run and transfer.
Find the right protein ladder
See the advantages of NuPAGE gels
We’ve saved the best for last…are you ready to take your western blots to the next level with fluorescent multiplexing? Recruit our easy-to-use iBright FL1000 Imaging System—designed to capture high-resolution, high-sensitivity fluorescent western blots—and our Alexa Fluor Plus conjugated secondary antibodies with enhanced sensitivity and low background. With this power combo, you can detect multiple proteins in the same blot.
Explore our range of chemiluminescent substrates
Get stunningly easy blot and gel documentation
For Research Use Only. Not for use in diagnostic procedures.