Bioinformatics designed for accuracy
Ion Reporter Software automates secondary and tertiary analysis for a variety of bioinformatics workflows, including variant calling, copy number detection, and fusion analysis. Its cloud-based scalability and customizable pipelines help to ensure reproducible results for any lab size, while built-in security and compliance features support a rigorous standard for data management.
The Ion Reporter data analysis process helps researchers get high quality data by prioritizing and annotating variants. It completes with a report that simplifies your bioinformatics path to discovery by enabling you to focus on finding the biological meaning of your data.
Ion Reporter data analysis process
Easy data analysis
Ion Reporter Software removes several barriers of executing bioinformatics workflows, enabling you to focus on the meaning of your data. Streamlined analysis with integrated run planning from Torrent Suite Software automates the primary analysis, data transfer, and secondary analysis in Ion Reporter Software.
Optimized workflows: designed to deliver consistent and accurate results
Ion Reporter Software analysis and variant calling helps you quickly focus on variants of interest supported by full annotation from public and proprietary databases.
Informatics for your application
Ion Reporter Software is designed with preconfigured and customizable workflows for a variety of research applications from inherited disease to reproductive health to oncology. Workflows are flexible to enable custom optimization and can also be locked to enable controlled use without depending on the advanced skills of a bioinformatician.
Ion Reporter XL Server System
The Ion Reporter XL Server System combines Ion Reporter Software with a robust computer server to provide a secure, integrated, NGS analysis solution within your own facility.
  • Accelerated performance—dual 20-core CPUs and 256 GB RAM enable fast data transfer and analysis for NGS workflows
  • Increased capacity—40 TB of usable storage offers adequate space for large projects and multi-sample studies
  • Secured environment—operate entirely within your laboratory network for compliant, controlled, and traceable analysis
Ordering information for Ion Reporter XL Server
Catalog # Name Size Price (USD) Qty
4487118XL Each
44,110.00
Resources
For Research Use Only. Not for use in diagnostic procedures.
PMR: 001137