Search
GeneArt Site-Directed Mutagenesis Service |
Site-directed mutagenesis is a molecular biology technique that is used to introduce specific changes at desired locations within a DNA fragment; with this technique, complete de novo synthesis is no longer required. Mutagenesis of existing genes can help you find answers for your research, including:
The GeneArt site-directed mutagenesis service introduces single or multiple mutations (substitutions, insertions, deletions, or truncations) into existing DNA sequences, quickly and economically. We use a targeted PCR-based process to introduce genetic variation precisely where you need it and then clone the construct into the vector of your choice. All final sequences are 100% sequence-verified and documented to help ensure that the DNA you receive is exactly what you’ve requested.
Download the Multiple Sequence Submission Form, enter your DNA sequence request(s) using Microsoft Excel software, and email it to our customer support team at geneartsupport@thermofisher.com.
For further information regarding this service or the order process, please contact geneartsupport@thermofisher.com
For your convenience, GeneArt mutagenesis services offers precise and customizable variant creation with guaranteed sequence verification and efficient processing times.
Variants ≤4 kb can be ordered directly via the GeneArt online order portal or emailgeneartsupport@thermofisher.com.
Variants >4 kb can be requested via the GeneArt online order portal or email but will require manual review before order placement.
Variants may contain up to four of the following modifications:
GeneArt mutagenesis service (Single variants: ≤5 variants from one template)
GeneArt mutagenesis 6+ service (Bundle variants: >5 variants from one template)
| Single variants | Business days |
|---|---|
| Up to 3 kb | 6 |
| >3 kb | 10 |
| Bundle variants | Business days |
| <10 variants | 7 |
| 10–20 variants | 10 |
| 21–48 variants | 13 |
| 49–96 variants | 16 |
| Additional 96 variants | +5 each |
| For very large projects (500 variants or more) | Please inquire |
Variants ≤4 kb can be ordered directly via the GeneArt online order portal or emailgeneartsupport@thermofisher.com.
Variants >4 kb can be requested via the GeneArt online order portal or email but will require manual review before order placement.
Variants may contain up to four of the following modifications:
GeneArt mutagenesis service (Single variants: ≤5 variants from one template)
GeneArt mutagenesis 6+ service (Bundle variants: >5 variants from one template)
| Single variants | Business days |
|---|---|
| Up to 3 kb | 6 |
| >3 kb | 10 |
| Bundle variants | Business days |
| <10 variants | 7 |
| 10–20 variants | 10 |
| 21–48 variants | 13 |
| 49–96 variants | 16 |
| Additional 96 variants | +5 each |
| For very large projects (500 variants or more) | Please inquire |