Search
Validated for a broad range of targets and samples according to ISO 16140-2:2016 (AFNOR validation) and AOAC certification requirements, the SureTect Assays provide the ultimate solution for all of your foodborne pathogen detection PCR testing requirements. Each SureTect PCR Assay contains all of the ready-to-use reagents and consumables for your food safety workflows:
Color-coded assay strip plates
Easy, safe separation of lysis and assay plates into 8-well strips
Cut strips to required number of tubes
Color-indicator in Proteinase K reagent
Efficient plate sealing
Pierceable lysis tube seals
SureTect PCR Assays are validated for specific foods and a broad range of samples according to ISO 16140-2:2016 (AFNOR VALIDATION) and/or AOAC and fully supported by our dedicated and experienced technical teams.
As the global food testing market continues to grow, food producers need faster, more efficient, and cost-effective solutions. That’s where alternative food testing approaches come in, offering rapid, reliable results that align with modern industry demands.
Our comprehensive eBook, "A guide to efficiently implementing a new, alternative food testing method in accordance with ISO 16140 – Part 3" is your roadmap to navigating, selecting, and verifying alternative workflows in line with an internationally recognized standard.
Follow the validation journey of a Thermo Scientific food pathogen detection solution through method validation according to ISO 16140-2:2016 and learn how Thermo Fisher can support your laboratory with the efficient and successful verification of an alternative method according to ISO 16140-part 3.
For Research Use Only. Not for use in diagnostic procedures.