Search
Relative quantitation and accurate identification of lipids from biological samples requires sophisticated software with a comprehensive database. Thermo Scientific LipidSearch software provides automated identification of lipids and integrates an entire dataset into a concise report showing statistical differences between sample groups.
| Automated lipid identification | Comprehensive database coverage |
|---|---|
| Unique peak detection algorithms are customized for LC-data dependent MS/MS including combining the information from CID/HCD fragmentations and MS2/MS3 spectra using the Thermo Scientific Orbitrap IQ-X and Tribrid mass spectrometers. Product ion search and annotation alignment algorithms are optimized for untargeted lipid identification. | The LipidSearch software database contains molecular lipid species (96 sub-classes) and their predicted fragment ions and provides the most complete coverage of different sample types including cells, human plasma, insect larvae, plants, seed oils, tissue and yeast. |
We’re here to help you maximize your free trial of LipidSearch software. Contact us if you would like more information on how to use it, or if you’d like to have a representative contact you.