Search
CaptureSelect ligands offer a unique affinity purification solution based on Camelid-derived single domain [VHH] antibody fragments. These small, 14 kD affinity ligands, are the platform solution for many biopharmaceutical purification challenge and have been proven for many applications to result in a higher yield and an increased purity of the protein of interest. In addition, CaptureSelect ligands can be tailored to protect the protein by applying mild elution conditions. The manufacturing in yeast, without animal derived components, results in a high quality product lacking toxins and viruses.
The proprietary CaptureSelect technology makes use of the uniqueness of single domain antibodies offering:
The development of any CaptureSelect ligand is based on the flexibility and specificity of the mammalian immune system, enabling ligands to be designed for almost any target and selected to have specific affinity, specificity and elution profiles.
The manufacturing process of the single domain antibodies involves a proprietary yeast system and is completely free of animal derived components. Life Technologies’ facility in The Netherlands includes reactors ranging from 10 L to 15,000 L reactors, micro- and ultra-filtration systems, and a clean room for final purification, allowing for rapid scale up and efficient industrial production of the affinity ligands.
Our production plant in Naarden, Netherlands has been built to an advanced engineering level, satisfying international standards for hygienic processing and for the handling of Genetically Modified Micro organisms (GMOs), and has been ISO 9001 certified since 1997.
| ![]() |
Interactive Decision Tool
Find your solution ›
Watch this webinar on novel antibody therapeutic purification
Check out the new Chromatography Learning Lab from Thermo Fisher Scientific, providing a robust range of webinars, articles, eBooks, infographics, and other digital resources to help you optimize the use of our purification products in your process.
Explore digital resource
For more information on CaptureSelect technologies
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
Pharmaceutical Grade Reagent. For Manufacturing and Laboratory Use Only.