Search
StemPro hESC SFM – a fully-defined, serum- and feeder-free medium (SFM) specially formulated for the growth and expansion of human embryonic stem cells (hESCs)
![]() | ![]() | ![]() | ![]() |
| SSEA4 staining of hESCs grown in STEMPRO® hESC SFM | Tra-1-81 staining of hESCs grown in STEMPRO® hESC SFM | BG02 cells grown in STEMPRO® hESC SFM | BG02 cells grown in STEMPRO® hESC SFM |
StemPro hESC SFM has been extensively tested and proven to maintain pluripotency in a growing list of hESC lines including:
| BG01 | BG02 | BG03 |
| H1 | H9 | BG01V |
| HUES9 | HUES6 | |
| CyT49 | Cyth203 | |
| Rhesus Monkey ES cells (line R366.4) | ||
Geltrex Reduced Growth Factor Basement Membrane Matrix is designed for use with STEMPRO hESC SFM as a matrix to support hESC growth. Geltrex has a consistently high lot to lot protein concentration and functional testing is performed on each lot.
For Research Use Only. Not for use in diagnostic procedures.