Search
Custom TaqMan® Small RNA Assays employ an innovative target-specific stem-loop reverse transcription primer to address the challenge of the short length of the small RNA. The primer extends the 3' end of the target to produce a template that can be used in standard TaqMan® Assay-based qPCR. Also, the stem-loop structure in the tail of the primer confers a key advantage if you are studying miRNAs: specific detection of the mature, biologically active miRNA.
Life Technologies offers a variety of kits for the isolation and analysis of miRNA and other small RNAs. Each is ideal for use in miRNA analysis because they enable quantitative recovery of small RNA (<200 nt) and maintenance of representative amounts of small RNA (eliminating experimental bias).
mirVana™ miRNA Isolation Kit | ||
RecoverAll™ Total Nucleic Acid Isolation Kit for FFPE | ||
TaqMan® MicroRNA Cells-to-CT™ Kit |
Two-step TaqMan® MicroRNA Assays, containing the TaqMan® Assay and reverse transcription primer specific for the target of interest, are designed for individual miRNA targets found in a wide variety of species including human, mouse, rat, Drosophila, C. elegans, and Arabidopsis. The TaqMan® MicroRNA Reverese Transcription Kit provides optimal TaqMan® MicroRNA Assay performance.
TaqMan® MicroRNA Reverse Transcription Kit | ||
Custom TaqMan® Small RNA Assays |
For the final quantitation step, the cDNA product generated in step 2 is mixed with TaqMan® Universal Master Mix II and added to the matched TaqMan® MicroRNA Assay.
Custom TaqMan® Small RNA Assays | ||
TaqMan® Universal Master Mix II |
Life Technologies offers a variety of kits for the isolation and analysis of miRNA and other small RNAs. Each is ideal for use in miRNA analysis because they enable quantitative recovery of small RNA (<200 nt) and maintenance of representative amounts of small RNA (eliminating experimental bias).
mirVana™ miRNA Isolation Kit | ||
RecoverAll™ Total Nucleic Acid Isolation Kit for FFPE | ||
TaqMan® MicroRNA Cells-to-CT™ Kit |
Two-step TaqMan® MicroRNA Assays, containing the TaqMan® Assay and reverse transcription primer specific for the target of interest, are designed for individual miRNA targets found in a wide variety of species including human, mouse, rat, Drosophila, C. elegans, and Arabidopsis. The TaqMan® MicroRNA Reverese Transcription Kit provides optimal TaqMan® MicroRNA Assay performance.
TaqMan® MicroRNA Reverse Transcription Kit | ||
Custom TaqMan® Small RNA Assays |
For the final quantitation step, the cDNA product generated in step 2 is mixed with TaqMan® Universal Master Mix II and added to the matched TaqMan® MicroRNA Assay.
Custom TaqMan® Small RNA Assays | ||
TaqMan® Universal Master Mix II |
Brochures
Product Information