Search
GeneArt Controlled Randomization Service |
At the forefront of genetic research and biopharmaceutical advancements is the creation and utilization of mutagenesis libraries. These libraries are powerful tools that enable scientists to explore gene function, engineer proteins, and develop therapeutics by introducing mutations into DNA sequences. Our GeneArt controlled randomization service is designed to provide high-quality, customizable mutagenesis libraries that meet the stringent demands of modern scientific research.
Applications of mutagenesis libraries are diverse and impactful, spanning multiple areas of scientific research and industrial use:
In a controlled randomization library, random nucleotides within a defined region will be mutated at a defined average mutation rate. GeneArt controlled randomization technology accurately directs the effective frequency of random nucleotide substitutions. This approach is designed to confine mutagenesis to specific regions or randomize the entire open reading frame. Randomized constructs are rigorously controlled to help ensure that the unchanged portions of the sequence are unaffected and that randomized portions contain the distribution of nucleotides that encode the desired amino acid composition.
All GeneArt controlled randomization products are sequenced and statistically analyzed to help ensure that they meet the following quality benchmarks:
Please download the questionnaire to submit project information. For secure data transfer, please login to the GeneArt Dashboard to access the file exchange.
For further information regarding this service or the order process, please contact geneartsupport@thermofisher.com.
| GeneArt ready-to-clone controlled randomization library |
|
|---|---|
| GeneArt cloned controlled randomization library |
|
Watch how-to videos and explainers on GeneArt Instant Designer.
Contact us
Request more information
North America and Asia: 1-800-955-6288
Ask for "GeneArt" when prompted
Europe: +49 (0)941 942 76-100
Order support:
Geneartsupport@thermofisher.com
Stay up to date on gene synthesis and protein synthesis service offerings.
Read white papers on de novo gene synthesis technologies.
![]()