Syringe Filters
Eliminate particles and microorganisms from your sample — reliably
Prevent the accumulation of fine particles that can block your chromatography column. Thermo Scientific Titan3, Target2, and Choice SyringeFilters remove interfering materials, fine particles, and microorganisms, providing you with cleaner sample extracts. These high-quality robust sample filtration solutions are available in a variety of syringe filter sizes, membranes, and housings for a wide range of samples and applications.
Contact us
Titan3, Target2, and Choice syringe filters
Syringe FilterApplication
Premium BrandRoutine BrandBasic Brand
Titan3 NylonTarget2 NylonChoice Nylon
  • General laboratory filtration
  • Filtration for most HPLC samples
Titan3 PTFE
(Hydrophobic)
Target2 PTFE
(Hydrophobic)
Choice PTFE
(Hydrophobic)
  • Filtration of aggressive organic, highly basic, or hot solutions, transducer protectors
  • Filtering aqueous solutions after prewetting with an alcohol
Titan3 PTFE
(Hydrophilic)
—Choice PTFE
(Hydrophilic)
  • Filtration of aggressive organic, highly basic, or hot solutions, transducer protectors
  • Filtering aqueous solutions no-prewetting required
Titan3 PVDF
(Hydrophobic)
Target2 PVDF
(Hydrophobic)
Choice PVDF
(Hydrophobic)
  • General biological filtration
  • Filtration of samples where high protein recovery is desired
——Choice PVDF
(Hydrophilic)
  • General biological filtration
  • Filtration of samples where high protein recovery is desired
  • Filtering aqueous solutions no-prewetting required
Titan3 Regenerated Cellulose (RC)Target2  Regenerated Cellulose (RC)Choice  Regenerated Cellulose (RC)
  • Low non-specific binding applications
  • Tissue culture media and general biological sample filtration
Titan3 PES
(Polyethersulfone)
Target2 PES
(Polyethersulfone)
Choice PES
(Polyethersulfone)
  • Ion chromatography
  • Tissue culture, proteins, and nucleic acids filtration
  • High-temperature liquids
Titan3 GMF
(Glass Microfiber)
Target2
(Glass Microfiber)
—
  • Dissolution testing
  • General filtration
Titan3 PolypropyleneTarget2 PolypropyleneChoice Polypropylene
  • Filtration of biological samples
  • Filtration of aggressive organic solutions
Titan3 Cellulose AcetateTarget2 Cellulose AcetateChoice Cellulose Acetate
  • Tissue culture media filtration, sensitive biological samples
What is the difference between Titan3, Target2, and Choice syringe filters?
 Titan3 Premium Syringe FiltersTarget2 Routine Syringe FiltersChoice Basic Syringe Filters
Range of filter sizes4mm to 30mm4mm to 30mm13mm to 25mm
High-quality filtration solutions   
Low extractable membranes and housing   
Plain polypropylene housing   
HPLC performance tested   
Enhanced Luer Lock inlet which provides secure attachment to syringes   
Low hold-up volume allows for the treatment of small sample volumes with minimal sample loss   
Color coded for easy identification and selection of the correct membrane and pore size   
30mm products pressure rated to 120psi for enhanced sample safety   
Most 30mm devices have a 1mm borosilicate glass pre-filter for increased filtrate volumes