Search
The Luminex 200 Instrument System sets the standard for multiplexing, offering the ability to perform up to 100 different tests in a single reaction volume on a flow cytometry–based platform. The Luminex 200 system is compatible with Invitrogen ProcartaPlex multiplex immunoassays and Invitrogen QuantiGene Plex multiplex gene expression assays.
Key features of the Luminex 200 Instrument Systems:
Multiplexing
Up to 80 analytes per sample using magnetic microspheres
Read time
96-well plate in ≤45 min (up to 12,800 tests/hour)
Dynamic range (typical)
≥3.5 logs
Daily start-up/shut down
≤30 min
|
|
Optics |
Lasers, APDs, PMTs |
Hardware |
Flow cytometry–based lasers, APDs, PMTs |
Footprint including PC (linear bench space) |
115 cm x 60 cm x 50 cm |
Weight (analyzer) |
49 kg (113 lb) |
Multiplex capacity |
Up to 100; (80 on MagPlex) |
Maximum available commercial assay plex |
80 |
Application |
Protein/nucleic acid |
Sample volume |
25–80 µL (assay dependent) |
Plate format |
96-well |
Read time |
45 min (96-well) |
Sensitivity (assay dependent) |
0.06–1 pg/mL (immunoassays) |
Reporter laser/optics |
532 nm (green) |
Dynamic range |
≥3.5 logs (RP1) |
Operational validation |
Calibration, verification, and fluidics |
21 CFR compliance |
Yes |
Automation |
Integration software |
Analytics (software) |
xPONENT basic plus Thermo Fisher Connect Platform |
For Research Use Only. Not for use in diagnostic procedures.