Validate the performance of particle counting and sizing instruments
Rely upon Thermo Scientific™ NIST Traceable Size Standards as a third party reference to calibrate and check performance when particle sizing instruments are used for solving real-world analytical problems. The Certificate of Analysis and Traceability to NIST helps in audits by answering the question: “How do you know the instrument is measuring properly?”
Key Applications
- Instrument calibration, development and testing of particle sizers, particle counters
- Instrument QC: check for instrument problems and shifts in laser function, check the calibration curve shape, etc.
- Equipment efficiency studies: vial cleaning, contamination studies, filter testing (including filter leaks), broad distribution material analysis
- Spacers: liquid crystal displays, precision microelectronics
Resources
CMD SchemaApp code