Search
The following plate readers have been determined to be compatible with the LanthaScreen Tb Assay. Click on a specific instrument to obtain an assay-specific setup guide, filter and optical settings, and available application notes and posters.
This technical note provides a method for verifying that a fluorescent plate reader is able to detect a change in time-resolved fluorescence energy transfer (TR-FRET) signal, confirming proper instrument set-up and a suitable response. The method is independent of any biological reaction or equilibrium and uses reagents that are on-hand for the LanthaScreen assay.
For Research Use Only. Not for use in diagnostic procedures.