LentiArray gRNAs
Invitrogen LentiArray CRISPR gRNA are pre-designed, pre-packaged gRNA lentiviruses designed for efficient gene knockout. LentiArray CRISPR gRNA designs have been created with a proprietary gRNA design algorithm and are optimized for maximum knockout efficiency without compromising specificity. LentiArray CRISPR gRNAs are available to support pre-screen assay development and post screen hit validation when performing high throughput screening experiments with the LentiArray CRISPR libraries. LentiArray CRISPR gRNA can also be used with LentiArray Cas9 lentivirus to perform genome editing experiments in hard-to-transfect cell lines and primary cells.
LentiArray CRISPR gRNA selection tool
Can't find what you are looking for? Contact us for more information about CRISPR products and services.
CRISPR Controls
High quality controls play an integral role in the successful development and performance of high throughput screens.  Don't forget to get your controls to help you optimize delivery conditions, maximize editing efficiency and establish hit selection criteria. 
Resources
  • Learning Center
    Access genome editing application resources for more success as you plan and execute your experiments.
  • CRISPR Validated Protocols
    Getting started with CRISPR? Need proven CRISPR validated protocols? Check out our collection of step-by-step validated protocols.
  • FAQs
    Find answers to everyday problems, we have consolidated a list of most commonly asked questions.
  • Webinar Series
    Master the art of CRISPR editing by tuning into our on-demand 3 part webinar series.
  • CRISPR Hands-on Workshop
    Our experienced team has designed a comprehensive four-day workshop to get you up to speed on CRISPR.
Support