Vanquish Duo UHPLC Systems
Double your throughput or deepen your sample knowledge using the Thermo Scientific Vanquish Duo UHPLC Systems, featuring a dual split sampler and two pumps, either as separate modules or combined in the dual pump. The Vanquish Duo UHPLC system has the same footprint as a single instrument, but contains two flow paths allowing you to run the same method on both flow paths or run two different methods simultaneously on the same instrument, improving your lab productivity, sample knowledge, and overall return on investment.
Highlights
Doubled productivity without sacrificing bench space
The Vanquish Duo UHPLC system provides two independent flow paths connected to two individual detectors. This unique system lets you run two analyses, either identical or not, on a single system to double throughput, accelerate method development, and improve sample characterization.
Complementary analysis
Analyze the same set of samples using multiple methods or column chemistries while reducing sample preparation time and costs and maximizing sample information for faster biopharmaceutical characterization, or simultaneous API quantification and impurity analysis for pharmaceuticals.
Innovative dual split sampler and dual pump
The Thermo Scientific Vanquish Dual Split Sampler includes two injection devices in a single housing, for two individually controlled channels. Additionally, the Thermo Scientific Vanquish Dual Pump has two built-in independent ternary pumps saving purchasing costs and reducing system height.
Versatility in analyte detection
A variety of detection options ensure you can collect the most information from your separation. All detector options offer excellent sensitivity, including diode array, fluorescence, or label-free detection of non-chromophoric substances via charged aerosol detection.
Biocompatible, easy and reliable tool-free connections
The fluidic pathway of the Vanquish Duo UHPLC system is fully biocompatible and fitted with Thermo Scientific Viper Fingertight Fittings, which support near-zero dead volume connections and enable sharper peaks and tool-free setup.
CDS software
The Vanquish Duo systems seamlessly integrate with our Thermo Scientific Chromeleon Chromatography Data System (CDS) to deliver superior compliance tools, networking capabilities, automation and data processing.
Featuring the ACT Ecolabel
Thermo Scientific Vanquish Duo UHPLC systems are certified with the prestigious My Green Lab® ACT® Ecolabel awarded by My Green Lab, Inc. The My Green Lab ACT Ecolabel serves as the premier ecolabel for lab products, encompassing consumables, chemicals, and equipment. Standing for Accountability, Consistency, and Transparency (ACT), this certification offers clear, standardized information about a product’s environmental impact across its life cycle covering manufacturing, energy and water use, packaging, and end-of-life disposal. Leverage the My Green Lab ACT Ecolabel to make informed purchasing choices based on transparent environmental data so you can advance sustainability without sacrificing performance.
What is Vanquish Duo?
Our Vanquish Duo UHPLC systems allow you to run two of the same or two different assays simultaneously.
The diagram below illustrates the time savings achieved when running two different assays on the Vanquish Duo simultaneously, in comparison to the time required to run the same two assays on a traditional single-channel HPLC system.
Video
System specifications
The table below provides an overview of the Vanquish Duo UHPLC systems specifications for a standard setup. Specifications may differ based on the specific system components selected. Your specifications may differ based on the components of your system. Find the specification sheets for the system components in the Component specifications, below.
Category Vanquish Duo UHPLC Systems
System pressure, maximum 1000 or 1500 bar, depends on configuration
Pump options (default gradient delay volume)
Binary (35 or 200 µL)
Quaternary (679 µL) 
Dual (679 µL)  
Delay volume dependent on pump(s) selected
Biocompatible Yes
pH range 2–12, chloride concentration up to 1 mol/L
System design Modular
Touchscreen user interface display compatible (optional) Yes, depends on system configuration
Solvent monitoring system compatible (optional) Yes, depends on system configuration
Customizable gradient delay volume to simplify method transfer Yes, with metering device
SmartInject technology for maximum injection precision and improved column lifetime
Yes
Injection volume range Standard: 0.01-25 µL 
Optional: 0.01-100 µL
Sample cooling
Yes, 4-40°C
Sample capacity (standard) 
Based on 12 mm OD vials (≤1.5 mL) for comparison
4 racks of 54 vials (≤ 1.5 mL) each, other rack formats available 
+ capacity of 12 × 22.5 mm OD vials (≤ 10 mL) in the carousel
Sampler extension available (optional)
Yes, with Vanquish Charger
Column compartment thermostatting
Yes, 5-120°C 
Still air and forced air
Column compartment pre-heater Active and passive 
Column compartment post-column cooler Yes
Compatible detectors
Vanquish Charged Aerosol Detector 
Vanquish Diode Array Detector  
Vanquish Fluorescence Detector 
Vanquish Multiple Wavelength Detector 
Vanquish Variable Wavelength Detector
Refractive Index Detector 
Mass Spectrometer
Component specifications
Vanquish Duo UHPLC systems are made up of the following components with detail available below.
  Vanquish Horizon Duo UHPLC System Vanquish Flex Duo UHPLC System
Pump
Download Specification Sheet
2 Vanquish Binary Pump H 2 Vanquish Binary Pump F 
or 
2 Vanquish Quaternary Pump F 
or 
1 Vanquish Dual Pump F
Autosampler
Download Specification Sheet
Vanquish Dual Split Sampler HT Vanquish Dual Split Sampler FT
Extended sample capacity (optional)
Download Specification Sheet
Vanquish Charger  (optional)
Column compartment
Download Specification Sheet
Vanquish Column Compartment H
Virtually zero dead volume system connections
Download Specification Sheet
Viper UHPLC Fittings Kits
Detectors
2 Detectors
Visit our HPLC and UHPLC Detectors page
Contact us to get a quote or discuss an HPLC/UHPLC solution
For Research Use Only. Not for use in diagnostic procedures.