Oligonucleotides custom-manufactured to your specifications. Choose from Applied Biosystems 5´ labeled fluorescent primers and primer pairs for use in fragment analysis.
Custom 5´ labeled primers are fluorescently labeled oligos with a choice of dye on the 5´ end. These products can be used for fragment analysis, gene expression studies, PCR, and genotyping research.
Features include:
  • Choice of delivered scales
  • All oligos are desalted. Larger scales of labeled primers have an option for HPLC purification
  • IMPORTANT: HPLC purification reduces the yield by half
Cat. No. Size HPLC yield 5′ Reporter dye Quantity Order
450007 10,000 pmol NA 6-FAM, TET, VIC, HEX, NED, PET 1 tube Order
450006 80,000 pmol 40,000 pmol minimum 6-FAM, TET, VIC, HEX, NED, PET 1 tube Order
450017 300,000 pmol 150,000 pmol minimum 6-FAM, TET, VIC, HEX, NED, PET 1 tube Order
Specifications
Format Delivered either in solution (either 1X TE or water) or dried down
Purification Desalted; option of HPLC purification with larger scale
Shelf life 12 months from manufacturing date
Shipping condition Room temperature
Storage condition –20°C
Custom 5´ labeled primer pairs include a fluorescently labeled oligo with a choice of dye on the 5´ end, along with an unlabeled oligo, in each pair. These products can be used in a variety of research applications.
Features include:
  • Choice of delivered scales
  • For use in fragment analysis, gene expression studies, genotyping, and PCR
  • All oligos are desalted
Cat. No. Size HPLC yield 5' Reporter dye Quantity Order
450056 10,000 pmol 5´ Labeled/Unlabeled Primer Pairs 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order
450059 80,000 pmol 5´ Labeled/Unlabeled Primer Pairs 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order
450062 300,000 pmol 5´ Labeled/Unlabeled Primer Pairs 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order
4304976 10,000 pmol 5´ Labeled/Unlabeled Di-Repeats 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order
4304977 80,000 pmol 5´Labeled/Unlabeled Di-Repeats 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order
4304978 300,000 pmol 5´ Labeled/Unlabeled Di-Repeats 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order
4304979 10,000 pmol 5´ Labeled/Unlabeled Di-Repeat + Tail 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order
4304981 80,000 pmol 5´ Labeled/Unlabeled Di-Repeat + Tail 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order
4304982 300,000 pmol 5´ Labeled/Unlabeled Di-Repeat + Tail 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order
Specifications
Format Delivered either in solution (either 1X TE or water) or dried down
Purification Desalted; option of HPLC purification with larger scale
Shelf life 12 months from manufacturing date
Shipping condition Room temperature
Storage condition –20°C
Support
  • Order support
    Contact order & web support, search FAQs, view order history, and track your order
  • Technical support
    Contact a technical application scientist, instrument maintenance & repair requests, and search product documents & FAQs