Microglia Differentiation Protocols
On this page:
Explore groundbreaking research on microglial differentiation with these three selected publications. Learn about microglia differentiation methods, access detailed workflows, and easily add necessary reagents to your cart to replicate these experiments. For precise protocols, consult specific scientific literature.
How to order
1. Select your protocol: Navigate to one of the three microglia differentiation protocols below.
2. Add to cart: Navigate to "Ordering information" below the process overview diagram, select quantity (1), and add products to your cart.
3. Proceed to checkout: Ensure all items are correct in your cart and proceed with the checkout process.
hPSC to microglia differentiation protocol (normoxic)
Publication: Guttikonda et al., Nat Neurosci, 2021
Description: Guttikonda et al., describes a method to generate microglia from human pluripotent stem cells (hPSCs) that offers a robust and efficient approach, enabling high purity and functionality of the derived microglia. This method does not require hypoxic conditions or sorting procedures. It is designed to produce microglia that closely resemble their primary counterparts, making it an invaluable tool for researchers working with microglia cultures.
Protocol highlights:
  • Reproducible: The protocol reproducibly generated a pure population of IBA1+ microglial cells out of 10 different hPSC lines.
  • Similarity to primary microglia: The hPSC-derived microglia express key microglial-specific markers (such as TMEM119, C1QA, CX3CR1, GPR34) at levels comparable to primary human microglia, ensuring functional relevance.
  • Efficient and timely production: The protocol allows for the efficient production of microglia within 25 days, making it faster than many existing methods.
  • Functional maturation: The protocol mimics in vivo conditions by co-culturing with cortical neurons and using cytokines like IL-34 and M-CSF, which are crucial for microglial survival and maturation.
  • Scalability: The method doesn't require sorting process. That would support the generation of large numbers of microglia, which is beneficial for extensive research and potential therapeutic applications.
  • In vitro modeling: The derived microglia are capable of performing key functions such as environmental surveying and phagocytosis, similar to their in vivo counterparts, making them suitable for in vitro disease modeling and drug testing.
Ensure you have all the necessary materials for the hPSC to microglia differentiation protocol (normoxic); products and their corresponding volumes have been pre-selected. Only one quantity of each product is needed to complete this workflow. See "Ordering information" below to add to cart.
Catalog # Name Size Price (USD) Qty
A1517001 Each
311.65
Online exclusive
332.00 Save 20.35
A1110501 Each
78.50
120-14E-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
120-05ET-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
A1516401 Each
280.00
100-18B-50UG 50 µg
193.65
Online exclusive
228.00 Save 34.35
100-20-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
300-07-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
200-06-20UG 20 µg
190.65
Online exclusive
224.00 Save 33.35
300-18-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
200-03-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
21103049 Each
106.65
Online exclusive
115.00 Save 8.35
17504044 Each
159.00
25030081 Each
39.65
Online exclusive
42.92 Save 3.27
450-02-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
450-10-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
200-34-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
300-25-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
12440053 Each
49.65
Online exclusive
55.50 Save 5.85
11765054 Each
40.65
Online exclusive
43.18 Save 2.53
hPSC to microglia differentiation protocol (hypoxic)
Publication: Ohtonen et al., Sci Rep, 2023
Description: The Ohtonen et al., (2023) protocol does not require a sorting procedure but requires hypoxic conditions. The protocol guides iPSCs through specific stages using defined cytokines and growth factors, and the resulting microglial-like cells are obtained without the need for sorting.
Protocol highlights:
  • No need for cell sorting: The protocol does not require cell sorting, simplifying the process and reducing the time and resources needed.
  • High yield of functional cells: The protocol results in a high proportion of functional microglia-like cells that respond to nucleotide stimulation.
  • Freezing capability: Microglial progenitors can be frozen at the D8 stage and later thawed for further differentiation, providing flexibility in experimental planning.
  • Reproducibility: The use of defined cytokines and growth factors ensures consistent and reproducible microglia differentiation outcomes.
  • Scalability: The protocol allows for the generation of large quantities of microglia-like cells, beneficial for extensive experimental setups.
  • Comprehensive functional assays: The protocol includes steps for functional assays such as Ca2+ transient measurements and phagocytosis, ensuring the generated cells are functionally relevant.
  • Versatility for disease modeling: It is effective in modeling neurodegenerative diseases, including Parkinson's disease, using iPSCs derived from patients with specific mutations.
  • Adaptability to different genetic backgrounds: The protocol accommodates iPSCs from different sexes and genetic backgrounds, allowing for the microglia culture and study of sex and genetic-specific effects on microglial function.
Ensure you have all the necessary materials for the hPSC to microglia differentiation protocol (hypoxic); products and their corresponding volumes have been pre-selected. Only one quantity of each product is needed to complete this workflow. See "Ordering information" below to add to cart.
Catalog # Name Size Price (USD) Qty
A1517001 Each
311.65
Online exclusive
332.00 Save 20.35
A1110501 Each
78.50
120-14E-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
A1516401 Each
280.00
11320033 Each
53.65
Online exclusive
55.50 Save 1.85
35050061 Each
58.25
15140122 Each
34.67
25080094 Each
18.65
Online exclusive
18.97
12585014 Each
76.25
100-18B-50UG 50 µg
193.65
Online exclusive
228.00 Save 34.35
100-20-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
300-18-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
300-07-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
200-06-20UG 20 µg
190.65
Online exclusive
224.00 Save 33.35
200-03-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
12440053 Each
49.65
Online exclusive
55.50 Save 5.85
A5669501 Each
97.92
Special offer
Online exclusive
Ends 15-Dec-2026
144.00 Save 46.08
300-25-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
200-34-50UG 50 µg
606.65
Online exclusive
714.00 Save 107.35
HSC to microglia differentiation protocol (normoxic, adapted)
Publication: Dorion et al., Mol Neurodegener, 2024
Description: The Dorion et al., (2024) microglia differentiation protocol is an improvement over the Blurton-Jones's lab publications (Abud et al., 2017, Mcquade et al., 2018), which is referred to as "2.0" in the publication. This method does not require hypoxic conditions or sorting procedures. The new protocol, named "2.9," offers several enhancements.
Protocol highlights:
  • Improved cell yield and adhesive properties: The 2.9 protocol results in a significant improvement in cell yield and adhesive properties compared to the 2.0 protocol.
  • Enhanced protein expression and functional competence: Cells derived using the 2.9 protocol show improved protein expression of select microglia markers, enhanced scavenging activities, and a better inflammatory response.
  • Closer resemblance to primary microglia: The transcriptomic profile of the resulting induced microglia (iMGLs) more closely resembles that of primary human microglia.
  • Simplified and cost-effective: The differentiation medium was simplified, requiring fewer reagents, which allows for a significant cost reduction. Additionally, the protocol does not require cell sorting or the use of a hypoxia chamber, making it more straightforward and accessible.
  • Functional validation in disease models: The protocol was validated by generating iMGLs from an ALSP patient carrying a pathogenic CSF1R variant. These cells showed functional alterations in migratory, phagocytic, and inflammatory activities, demonstrating the protocol's utility in microglia culture experiments studying disease-related microglial dysfunction.
Ensure you have all the necessary materials for the HSC to microglia differentiation protocol (normoxic, adapted); products and their corresponding volumes have been pre-selected. Only one quantity of each product is needed to complete this workflow. See "Ordering information" below to add to cart.
Catalog # Name Size Price (USD) Qty
12571063 Each
61.65
Online exclusive
67.25 Save 5.60
35050061 Each
58.25
17504044 Each
159.00
41400045 Each
87.50
15140122 Each
34.67
200-34-50UG 50 µg
606.65
Online exclusive
714.00 Save 107.35
100-21-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
300-25-10UG 10 µg
190.65
Online exclusive
224.00 Save 33.35
300-03-5UG 5 µg
78.65
Online exclusive
92.25 Save 13.60
300-31-20UG 20 µg
190.65
Online exclusive
224.00 Save 33.35
Intended use statements may vary.