Search
Flexible tools for TR-FRET assay development:
Whether you are routinely running assays in-house to support discovery programs, or need generic reagents to quickly build new TR-FRET assays, our portfolio of LanthaScreen reagents can enable your assay development program. Our LanthaScreen toolbox offers a collection of Terbium and Europium reagents for a true 'mix and match' experience. Our Europium labeled antibodies are not only compatible with fluorophores from Life Technologies (such as Alexa Fluor 633 and 647), but are compatible with other red fluorophores such as CyDyes and ULight dyes.
Choose from:
If you don't see your antibody of interest on our list, let us label your antibody for you.
Inquire about our custom labeling services ›
Epitope tags facilitate the purification of proteins for use in many applications. Such tags can also be used to indirectly label proteins through the use of LanthaScreen anti-epitope tag antibodies.
Indirect labeling of a protein through the use of an anti-epitope tag antibody can be advantageous when working with partially purified protein, or in cases in which the direct labeling of the protein would negatively impact protein activity.
Our products are available as kits or stand alone antibodies. Trial kits contain 100mL of TR-FRET dilution buffer (PV3574), which is recommended for antibody dilution, and a positive control peptide for the antibody when available. Europium antibodies are compatible with fluorophores purchased from Life Technologies or other suppliers including but not limited to Alexa Fluor 647, Cy3, Cy5, and ULight dyes. Terbium antibodies are compatible with fluorescein or GFP.
| Product | Label | Trial Pack | 25ug | 1mg |
| LanthaScreen Eu-anti-DYKDDDDK | Europium | PV6034 | PV6026 | PV6027 |
| LanthaScreen Eu-anti-GST | Europium | PV5594 | PV5595 | |
| LanthaScreen Eu -anti-His | Europium | PV5596 | PV5597 | |
| LanthaScreen Eu-anti-GFP | Europium | A14173 | A14190 | |
| LanthaScreen Tb-anti-GST | Terbium | PV3550 | PV3551 | |
| LanthaScreen Elite Tb-anti-His | Terbium | PV5863 | PV5895 | |
| LanthaScreen Tb-anti-GFP | Terbium | A13391 | A13392 |
Our secondary anti-species antibodies bind to their corresponding species IgG with low nanomolar apparent Kds, and show negligible cross-reactivity with other species. These antibodies can be used to rapidly develop assays when an anti-target antibody is already available. The labeled anti-species antibody will bind to the anti-target IgG completing the TR-FRET pair for easy assay development.
Our products are available as kits or stand alone antibodies. Kits contain a positive control peptide for the antibody, and 100 mL of TR-FRET dilution buffer (PV3574) as a dilutent for the antibody during biochemical assay development.
| Product | Label | Trial kit (10ug) | Trial kit (50ug) | 50ug Streptavidin only | 1mg Streptavidin only |
| LanthaScreen Eu-Streptavidin | Europium | PV6030 | PV5899 | PV6025 | |
| LanthaScreen Tb-Streptavidin | Terbium | PV3965 | PV3965 | PV3966 |
Life Technologies offers do-it-yourself reagents or a custom service for labeling of your antibody or protein of interest. Reagents include amine-reactive, isothiocyanate-functionalized LanthaScreen chelates and thiol-reactive, maleimide-functionalized LanthaScreen terbium chelates for development of TR-FRET assays in-house. Europium chelate reagents are available on a custom basis.
Alternatively, we can custom label your antibody (or other protein) with either a europium or terbium chelate. Let us know what antibody you would like us to label (vendor information) or supply antibody in a buffer without carrier protein (i.e. BSA), and we will label it for you.
To learn more about custom labeling and gain access, submit an online inquiry
More information | Related products |
For Research Use Only. Not for use in diagnostic procedures.