Search
High Molecular Weight (HMW) DNA Extraction |
High Molecular Weight (HMW) DNA consists of long, intact DNA molecules, important for long-read sequencing analysis in structural variation studies. Reliable isolation methods for HMW DNA are vital to minimize fragmentation, help ensure purity, and maximize yield, all important for accurate genomic analyses. This can be particularly crucial in oncology research where HMW DNA analysis is used for detecting structural variants and characterizing tumor heterogeneity in solid tumor and liquid biopsy samples.
To help advance comprehensive genomic analysis including long-read sequencing applications, the Applied Biosystems MagMAX HMW DNA Kit has been designed to isolate HMW DNA with large intact fragments utilizing Dynabeads magnetic bead technology and optimized chemistries along with automated and manual workflows to enhance efficiency, reproducibility, and productivity. High-quality large genomic DNA can be achieved consistently, helping provide accurate and reliable long-read sequencing analyses to help drive genomic studies and cancer research efforts forward.
Long-read sequencing technologies, such as those developed by Pacific Biosciences (PacBio) and Oxford Nanopore Technologies, offer the ability to read sequences of DNA that are thousands of base pairs long. This contrasts with short-read sequencing technologies, which typically read DNA sequences that are only a few hundred base pairs long. The longer reads provided by long read sequencing are particularly beneficial for:
Obtaining high-quality HMW DNA can be difficult due to the large fragment size as well as added complications that can arise from common sample types and existing isolation methods. Technical challenges scientists face require reliable methods and optimized protocols that aim to address common challenges such as:
Reliable preparation of HMW DNA is important for the success of long-read sequencing analysis. Effective isolation methods enable long-read sequencing by addressing common challenges associated with HMW DNA isolation and meeting workflow needs associated with common techniques, including:
Compare the advantages of long-read sequencing and short-read sequencing techniques. Learn about common challenges with HMW DNA isolation and how to solve them for improved extraction quality, efficiency, and performance in downstream applications.
Leveraging Dynabeads magnetic bead-based technology, the MagMAX HMW DNA Kit enables effective HMW DNA isolation by helping provide:
Designed for long-read sequencing applications, the MagMAX HMW DNA Kit isolates high-integrity DNA with large intact fragments from blood, cell, and tissue samples with optimal efficiency and reproducibility. Suitable for both manual and automated workflows, the kit supports diverse needs and lab scalability.
The MagMAX HMW DNA Kit leverages the MagMAX DNA Multi-Sample Ultra 2.0 Kit chemistry with an added booster reagent and optimized workflow for HMW DNA extraction. If you are using the DNA Multi-sample Ultra 2.0 Kit for genomic DNA extraction, you just need the additional booster along with optimized protocols to target HMW DNA fragments.
Enhance the efficiency of your HMW DNA extraction workflow using the MagMAX HMW DNA Kit on Thermo Scientific KingFisher instruments. Automated protocols are available for Duo Prime, Flex, and Apex instrument models.
Figure 1. HMW DNA isolation from whole blood, cells, or tissue samples using the MagMAX HMW DNA Kit. Extract DNA manually with DynaMag magnetic separation racks or automate the workflow using a KingFisher Duo Prime, KingFisher Flex, or KingFisher Apex purification system.
Compare the performance of on market HMW DNA isolation kits in terms of DNA integrity, yield, and fragment size distribution.
Figure 2. Average DNA yields isolated from cells, fresh and frozen whole blood, and tissue using MagMAX HMW DNA Kit. Average DNA yields obtained from HMW genomic DNA isolated from cells (1 x 106 input), fresh and frozen whole blood (200 µL input), and tissues (8 mg brain, 2 mg tissue) with MagMAX HMW Kit on KingFisher purification instruments. Yield was obtained using Qubit technologies and chemistries for broad range analysis. Average yields observed across all samples were greater than 3 µg. Yields vary from sample to sample and donor to donor.
| Sample type | Source | Average yield (µg) | Average 260/280 | Average 260/230 |
|---|---|---|---|---|
| Cells | HeLa | 11.24 | 1.87 | 2.29 |
| Whole blood | Fresh | 6.49 | 1.88 | 2.22 |
| Frozen | 6.42 | 1.88 | 2.19 | |
| Tissue | Brain | 7.25 | 1.88 | 2.18 |
| Spleen | 7.34 | 1.92 | 2.33 |
Table 1. Purity and quality performance using NanoDrop instrument technologies to obtain average absorbance ratios of 260/280 and 260/230 from HMW genomic DNA isolated from cells (1 x 106 input), fresh and frozen whole blood (200 µL input), and tissues (8 mg brain, 2 mg tissue) with MagMAX HMW Kit on KingFisher purification instruments.
Figure 3. Average DNA fragment size isolated from cells, fresh and frozen whole blood, and tissues using MagMAX HMW DNA Kit. Largest genomic size obtained from averaged HMW genomic DNA isolated from cells (1 x 106 input), fresh and frozen whole blood (200 µL input), and tissues (8 mg brain, 2 mg tissue) with MagMAX HMW Kit on KingFisher purification instruments. Sizing was analyzed using Agilent Femto Pulse instrumentation and chemistries.
Figure 4. Percent DNA fragment recovery ≥40 kb from cells, fresh and frozen whole blood, and tissues using MagMAX HMW DNA Kit. Percent fragment recovery greater than or equal to 40 kb from genomic DNA isolated from cells (1 x 106 input), fresh and frozen whole blood (200 µL input), and tissues (8 mg brain, 2 mg tissue) with MagMAX HMW Kit on KingFisher purification instruments. Sizing was analyzed using Agilent Femto Pulse instrumentation and chemistries.
Discover how our HMW DNA isolation solution is transforming long-read sequencing applications through in-field studies and testimonials from our customers.
“You have no competition, don’t bother with other kits.”
Ming Ta Michael Lee, PhD, VP of Biobank Operations and Logistics, Galatea Bio, Inc
“The kit is simple, quick, and scalable. You can be confident that it will yield high-quality high molecular weight DNA from your samples.”
Alexis Tapanes-Castillo, PhD Associate Professor for College of Health Sciences & Technology at St. Thomas University
Long-read sequencing applications help provide critical insights into cancer biology, identifying a wider range of genomic variation that shorter reads can miss. This more complete picture of genomic variation offers a more comprehensive understanding of cancer-causing mutations that could ultimately transform the development of cancer diagnostics and targeted therapeutics.
Key long-read sequencing techniques in cancer genomics:
The new HMW DNA isolation kit supports these applications, offering high-quality intact DNA fragments essential for advancing cancer genomics research. Reliable tools like this kit are crucial for enhancing diagnostic accuracy and treatment efficacy in cancer research.
When applied to hemato-oncology, long-read sequencing can offer a more complete and accurate view of the genomic, transcriptomic, and epigenetic alterations in hematological cancers. Longer reads provide improved structural variant (SV) detection, enabling comprehensive identification of complex genomic alterations such as large insertions, deletions, inversions, and translocations. Outside of HMW DNA analysis, long-read sequencing can also enhance the understanding of gene fusions and isoforms by sequencing full-length RNA transcripts which is important for diagnosing and targeting therapies in leukemia. Long-read sequencing platforms can also directly detect epigenetic modifications, such as DNA methylation, offering valuable insights into the epigenetic landscape of hematological cancers.
Interested in hematological cancer research?
Learn more about upstream nucleic acid isolation from blood and bone marrow samples using the MagMAX Sequential DNA/RNA Kit
Long-read sequencing also allows for long-range phasing, crucial for understanding allele-specific gene expression and the impact of mutations on oncogenes and tumor suppressor genes. This can be applied to single cell isolation and analysis of rare cells, such as circulating tumor cells (CTCs), found in blood, uncovering their unique genetic and transcriptomic profiles. By identifying genomic mutations and alterations in circulating cells, the development of personalized cancer treatment strategies for targeting hematological malignancies is made possible.
Learn more about cell isolation using Invitrogen Dynabeads magnetic beads
CTCs are a component of liquid biopsies, which involve analyzing blood or other biofluids to detect and analyze circulating biomarkers. The comprehensive analysis liquid biopsy can provide contributes to an overall deeper understanding of cancer biology, facilitating in the development of personalized therapies and treatment strategies, enabling further advancements in the oncology field overall.
Want to learn more about cancer research through the lens of genetic and protein biomarkers?
Download our eBook: Synergetic biomarkers: Fueling advancements in cancer research
Effective isolation of smaller genomic DNA (gDNA) fragments is important for genomic studies outside of long read sequencing. These smaller fragment targets are often used in applications such as next generation sequencing, dPCR, and other molecular biology techniques. Reliable analyses often rely on the isolation of the DNA. MagMAX automated magnetic bead-based kits can support this upstream sample preparation with reliable and efficient extraction methods.
Optimized for high-quality isolation of genomic DNA from a wide array of sample types with manual and automated protocols to fit flexible workflow needs. Standalone reagents available for scale-up and customizability.
| Product | MagMAX HMW DNA Kit | MagMAX DNA Multi-Sample Ultra 2.0 Kit | MagMAX DNA Multi-Sample Ultra Kit |
|---|---|---|---|
![]() | ![]() | ![]() | |
| Qualified sample types | Blood, cells, tissues | Blood, cells, tissues, bone marrow, saliva, buccal swabs, buffy coat, blood cards | Blood, cells, tissues, buccal swabs, saliva, blood cards, urine |
| DNA fragment size range | 100–300 kb | 1–40 kb | 1–40 kb |
| KingFisher compatibility | Duo Prime, Flex, Apex | Duo Prime, Flex, Apex | Duo Prime, Flex |
| Standalone reagents available | Yes | Yes | No |
| Downstream applications | Long-read sequencing, microarray | PCR, NGS, microarray | PCR, NGS |
| Cat. No. unit size | A36626, 100 preps A36627, 100 preps* | A36570, 1 kit | A25597, 500 preps A25598, 2,500 preps A45721, 1 kit* |
*with cell and tissue extraction buffer
As a leading supplier of world-class reagents and instrumentation, we also offer tailored manufacturing solutions for companies developing next-generation products and services. Our experienced OEM and commercial supply team is prepared to support your specific needs, from concept to commercialization.
For Research Use Only. Not for use in diagnostic procedures.