Search
Join the conversation
Primary XPS region: Al2p
Overlapping regions: Pt4f, Cu3p
Binding energies of common chemical states:
| Chemical state | Binding energy Al2p |
|---|---|
| Al metal | 72.6 eV |
| Aluminosilicate | 74.4 eV |
| Al oxide | 74.6 eV |
| Al oxide on Al foil | 75.6 eV |
Oxide/foil referenced to Al2p metal peak.
Silicate and alumina referenced to adventitious C1s peak at 284.8eV.
Experimental Information
Interpretation of XPS Spectra
Symbol: Al
Date of discovery: 1825
Name origin: Latin alumen
Appearance: silver
Discoverer: Hans Christian Ørsted
Obtained from: bauxite
Melting point: 933.52 K
Boiling point: 2740.15 K
Density[kg/m3]: 2700
Molar volume: 10.00 × 10-6 m3/mol
Protons/Electrons: 13
Neutrons: 14
Shell structure: 2,8,3
Electron configuration: [Ne]3s23p1
Oxidation state: 3
Crystal structure: cubic face centered
While ancient civilizations used aluminum salts for dyeing mordants and astringents, it wasn’t until 1825 that H. Ørsted isolated aluminum. The biggest breakthrough came in 1886, when C. Hall developed an inexpensive electrolytic process for extracting aluminum from the ore bauxite. This process is still commonly used today. Aluminum is valued for its light weight, strength, durability, and resistance to oxidation, and has propelled the use of aluminum alloys for aircraft and rockets. Evidence exists that high levels of aluminum may be toxic, although aluminum is generally considered less toxic than most heavy metals. Although the durability of aluminum is well-known, certain chemicals, such as mercury, can cause corrosion in aluminum. This is the reason why mercury thermometers are not allowed on aircrafts.
Products
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.