Consistent and reliable reagents for diagnostic testing, development, and biopharma manufacturing quality control, and clinical trial assay development
Molecular diagnostics testing, assay development, and biopharma processes require real-time PCR (qPCR) reagents with the highest level of consistency and reliability. With stringent cGMP manufacturing, excellent performance, and compliance documentation support, Applied Biosystems TaqPath general purpose reagents are designed to deliver confidence and reliability for even your most demanding applications. TaqPath master mixes are labeled “For Laboratory Use” and optimized for exceptional lot-to-lot consistency. 
For master mixes that support research workflows, see our TaqMan qPCR master mixes.
TaqPath master mixes are optimized to simplify your IVD assay development and regulatory submissions, clinical trials QC, etc.  Supporting compliance package documents are available on the product pages accessed below.  
Which TaqPath master mix is right for your application?
TaqPath DuraPlex 1-Step RT-qPCR Master Mix
TaqPath BactoPure Microbial Detection Master Mix
TaqPath ProAmp Master Mix
TaqPath qPCR Master Mix, CG
Check price or
order now
Check price or
order now
Check price or
order now
Check price or
order now
Recommended applicationsPathogen detection
Gene expression
Microbial detectionSNP genotyping
Copy number variation
Gene expression
MicroRNA analysis
Target
RNA
DNA
gDNA
cDNA
Concentration
4X
2X
2X
2X
Multiplexing and passive reference dye
Available in two versions:
With ROX (up to 5 targets)
No ROX (up to 6 targets)
Available in two versions:
With ROX (up to 3 targets)
No ROX (>3 targets)
Available in two versions:
With ROX (up to 3 targets)
With Mustang Purple (>3 targets) 
With ROX (up to 3 targets)
Reverse transcriptase
Thermostable MMLV RT included
N/A
N/A
Purchase RT separately and use before qPCR
Cycling mode/run timeStandard: ~110 min
Fast: ~60 min
Standard: ~120 min
Fast: ~50 min
Standard: ~120 min
Fast: ~50 min
Standard: ~50 min
Fast: ~40 min
Carryover contamination
UNG compatible (UNG not included)
With UNG
With UNG
With UNG
Assay recommendation
TaqPath qPCR assays
TaqPath qPCR assays
TaqPath qPCR assays
TaqPath qPCR assays
Request a sample
Request a sample
Request a sample
Request a sample
TaqPath master mixes are general purpose reagents labeled ‘For Laboratory Use’ and manufactured in an ISO 13485-certified and FDA-registered facility that adheres to cGMP principles. Every lot is functionally tested to help ensure lot-to-lot reproducibility for Ct consistency across a wide dynamic range.
For additional workflow components that support biopharma applications, see our QualTrak qPCR biopharma ecosystem.
Optimize your multiplexing with success, download resources ›
  TaqMan multiplexing—solutions for efficient multiplex PCR