Search
The small footprint design of the Sorvall WX+ ultracentrifuge series helps maximize space in your lab. Speeds of up to 100,000 rpm match your lab’s needs and provide high performance for applications from genomics to proteomics. An open rotor system lets you use most ultraspeed rotors from other major manufacturers for flexibility.
The Sorvall MTX/MX+ micro-ultracentrifuge series, which produces over 1 million x g, features advanced functionality in a compact footprint, with both floor-standing and benchtop configurations. Quiet operation, as low as 45 dB, lets technicians work with minimal distraction. The high torque drive in the MTX 150 centrifuge provides fast acceleration and deceleration rates, which means quick separations and less waiting at the end of the run.
Lightweight Fiberlite carbon fiber rotors are versatile options for ultracentrifuges. These fatigue-resistant rotors do not need to be derated over time. Rugged titanium rotors come in a variety of speed and capacities to accommodate different tubes. Continuous flow and zonal rotors are also available for high volume downstream processing, including density gradient separations.
| Sorvall WX+ Series features |
![]() |
| Sorvall MTX and MX+ Series features |
![]() |
Ultracentrifuges |
Micro-ultracentrifuges |
| (maximum capacity 6x250 mL) | (maximum capacity 6x 30 mL) |
Sorvall WX 100+ (up to 100,000 rpm) |
Sorvall MX120 Plus |
Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.