Search
Silencer Select siRNAs |
By using our Silencer Select siRNA, you can achieve clean, consistent phenotypic data, helping lead to more accurate and reproducible experimental outcomes. Our Silencer Select siRNAs are:
Explore Silencer Select siRNAs for lncRNA or search below for your lncRNA gene of interest.
Click on the tabs below to view data on targeted gene silencing with Silencer Select siRNAs.
At Thermo Fisher Scientific, we combined a powerful machine learning method with performance data from thousands of siRNAs to better understand the link between the sequence of an siRNA and its thermodynamic properties, target location, and silencing efficiency [1]. This is unlike most siRNA design algorithms which predict effective siRNAs that induce 70% target mRNA knockdown with only ~80% confidence and are inadequate for predicting more efficient siRNAs. RNAi applications demand better efficiency, leading us to build the Silencer Select algorithm and validate its functionality.
Silencer Select algorithm verification demonstrated our algorithm has:
Figure 1. Silencer Select siRNA design algorithm significantly improves effective siRNA prediction accuracy. Silencer Select siRNAs consistently outperformed those designed with a previous algorithm, delivering stronger mRNA knockdown across 40 gene targets in HeLa cells. The inset shows percentage of siRNAs that achieved ≥70% and ≥80% knockdown, highlighting improved potency and design accuracy.
Sequence-specific off-target effects are one of the primary reasons for false positive results in RNAi experiments. In addition to the potency improvements afforded by the Silencer Select algorithm, advanced bioinformatic filtering criteria removes off-targeting due to sequence. Combined with incorporation of novel chemical modifications demonstrated to improve siRNA specificity, Silencer Select siRNAs are highly potent with low off-target effects.
Although using siRNAs at low concentrations decreases off-target effects, further specificity gains can be made using bioinformatic filtering to predict and eliminate potentially inefficient siRNAs.
Our Silencer Select siRNA design process helps minimize off-target effects via:
Silencer Select siRNAs incorporate Locked Nucleic Acid (LNA) modifications to enhance their stability, binding affinity, and specificity. LNA-modified bases have the ribose ring chemically "locked" by a methylene bridge connecting the 2'-oxygen and the 4'-carbon. Adding LNA modifications to Silencer Select siRNA leads to:
Figure 3. Silencer Select siRNA modifications reduce off-target effects and yield more reliable phenotypic data. Modified siRNAs preserved expected mitosis and apoptosis phenotypes for PLK and WEE1. In contrast the off-target apoptotic phenotypes elicited by 10 unmodified siRNAs were eliminated with addition of the Silencer Select modifications-demonstrating the modifications support generation of more reliable data.
siRNA mediated gene silencing enables the study of gene function with exceptional power and ease. However, currently available siRNAs for the same target do not always produce the same phenotype due to a combination of inconsistent silencing and sequence-specific off-target effects. Silencer Select siRNAs are designed with modifications that help overcome these issues, leading to more reliable data
Multiple publications confirm that higher siRNA concentrations lead to increased off-target effects [2,3]. This insight drove our efforts to design more potent siRNAs that can be used at lower concentrations, by significantly enhancing our siRNA design algorithms.
Silencer Select siRNAs
Figure 5.Silencer Select siRNAs offer up to 100X higher potency compared to other siRNAs. Comparison of target knockdown in HeLa cells using Silencer Select siRNAs and siRNAs from two other suppliers to the same 10 different targets demonstrates the superior potency of Silencer Select siRNAs.
At Thermo Fisher Scientific, we guarantee that when you purchase two Invitrogen Silencer Select Pre-designed siRNA to the same target, those two siRNAs will silence the target mRNA by 70% or more. To qualify for the guarantee, siRNAs must have been transfected at ≥5 nM with proper controls and mRNA levels detected 48 hours post-transfection. Follow the link to learn more about our industry-leading guarantee.
A number of Invitrogen siRNAs targeting common human targets have been functionally tested by Thermo Fisher Scientific scientists and verified to reduce target mRNA levels by 70% or greater. The data sheet included with each of these siRNAs shows the extent of mRNA knockdown observed during testing and the exon targeted by the siRNA. We offer full sequences for predesigned and validated siRNAs with purchase.
At Thermo Fisher Scientific, we combined a powerful machine learning method with performance data from thousands of siRNAs to better understand the link between the sequence of an siRNA and its thermodynamic properties, target location, and silencing efficiency [1]. This is unlike most siRNA design algorithms which predict effective siRNAs that induce 70% target mRNA knockdown with only ~80% confidence and are inadequate for predicting more efficient siRNAs. RNAi applications demand better efficiency, leading us to build the Silencer Select algorithm and validate its functionality.
Silencer Select algorithm verification demonstrated our algorithm has:
Figure 1. Silencer Select siRNA design algorithm significantly improves effective siRNA prediction accuracy. Silencer Select siRNAs consistently outperformed those designed with a previous algorithm, delivering stronger mRNA knockdown across 40 gene targets in HeLa cells. The inset shows percentage of siRNAs that achieved ≥70% and ≥80% knockdown, highlighting improved potency and design accuracy.
Sequence-specific off-target effects are one of the primary reasons for false positive results in RNAi experiments. In addition to the potency improvements afforded by the Silencer Select algorithm, advanced bioinformatic filtering criteria removes off-targeting due to sequence. Combined with incorporation of novel chemical modifications demonstrated to improve siRNA specificity, Silencer Select siRNAs are highly potent with low off-target effects.
Although using siRNAs at low concentrations decreases off-target effects, further specificity gains can be made using bioinformatic filtering to predict and eliminate potentially inefficient siRNAs.
Our Silencer Select siRNA design process helps minimize off-target effects via:
Silencer Select siRNAs incorporate Locked Nucleic Acid (LNA) modifications to enhance their stability, binding affinity, and specificity. LNA-modified bases have the ribose ring chemically "locked" by a methylene bridge connecting the 2'-oxygen and the 4'-carbon. Adding LNA modifications to Silencer Select siRNA leads to:
Figure 3. Silencer Select siRNA modifications reduce off-target effects and yield more reliable phenotypic data. Modified siRNAs preserved expected mitosis and apoptosis phenotypes for PLK and WEE1. In contrast the off-target apoptotic phenotypes elicited by 10 unmodified siRNAs were eliminated with addition of the Silencer Select modifications-demonstrating the modifications support generation of more reliable data.
siRNA mediated gene silencing enables the study of gene function with exceptional power and ease. However, currently available siRNAs for the same target do not always produce the same phenotype due to a combination of inconsistent silencing and sequence-specific off-target effects. Silencer Select siRNAs are designed with modifications that help overcome these issues, leading to more reliable data
Multiple publications confirm that higher siRNA concentrations lead to increased off-target effects [2,3]. This insight drove our efforts to design more potent siRNAs that can be used at lower concentrations, by significantly enhancing our siRNA design algorithms.
Silencer Select siRNAs
Figure 5.Silencer Select siRNAs offer up to 100X higher potency compared to other siRNAs. Comparison of target knockdown in HeLa cells using Silencer Select siRNAs and siRNAs from two other suppliers to the same 10 different targets demonstrates the superior potency of Silencer Select siRNAs.
At Thermo Fisher Scientific, we guarantee that when you purchase two Invitrogen Silencer Select Pre-designed siRNA to the same target, those two siRNAs will silence the target mRNA by 70% or more. To qualify for the guarantee, siRNAs must have been transfected at ≥5 nM with proper controls and mRNA levels detected 48 hours post-transfection. Follow the link to learn more about our industry-leading guarantee.
A number of Invitrogen siRNAs targeting common human targets have been functionally tested by Thermo Fisher Scientific scientists and verified to reduce target mRNA levels by 70% or greater. The data sheet included with each of these siRNAs shows the extent of mRNA knockdown observed during testing and the exon targeted by the siRNA. We offer full sequences for predesigned and validated siRNAs with purchase.
Long non-coding RNAs (lncRNAs) are a type of non-coding RNAs (ncRNAs), typically over 200 nt, that are abundant in the mammalian transcriptome.
lncRNAs typically have lower expression levels compared to coding genes and expression can vary across cell lines. Improve results for knockdown of lncRNA with these suggestions:
Results of targeted knockdown in HeLa and HEK293T cells indicate that two or more siRNA produced at least 50% knockdown. 80% knockdown or more was obtained for six out of the seven targets. Knockdown was effective for lncRNA with nuclear or cytoplasmic localization.
Figure 6. lncRNA produces effective knockdown across different targets. siRNA transfection of HeLa and HEK293T cells highlights the exceptional performance of Silencer Select lncRNA to knockdown expression across different gene targets.
Transfection reagents alone or in combination with siRNAs may lead to a nonspecific decrease in cell viability. This may impact interpretation of knockdown results. Five different siRNA against seven different lncRNA were transfected into the indicated cells. No significant decrease in viability was detected.
Figure 7. Cells transfected with siRNA retain high viability. Cell viability data demonstrates the low toxicity of different lncRNA transfected into HeLa and HEK293T cells.
lncRNAs typically have lower expression levels compared to coding genes and expression can vary across cell lines. Improve results for knockdown of lncRNA with these suggestions:
Results of targeted knockdown in HeLa and HEK293T cells indicate that two or more siRNA produced at least 50% knockdown. 80% knockdown or more was obtained for six out of the seven targets. Knockdown was effective for lncRNA with nuclear or cytoplasmic localization.
Figure 6. lncRNA produces effective knockdown across different targets. siRNA transfection of HeLa and HEK293T cells highlights the exceptional performance of Silencer Select lncRNA to knockdown expression across different gene targets.
Transfection reagents alone or in combination with siRNAs may lead to a nonspecific decrease in cell viability. This may impact interpretation of knockdown results. Five different siRNA against seven different lncRNA were transfected into the indicated cells. No significant decrease in viability was detected.
Figure 7. Cells transfected with siRNA retain high viability. Cell viability data demonstrates the low toxicity of different lncRNA transfected into HeLa and HEK293T cells.
We synthesize and purify each siRNA in an advanced facility that is ISO13485 and ISO9001 certified to meet the highest quality standards. As part of our rigorous quality control procedures, each RNA oligonucleotide is analyzed by LCMS and/or analytical HPLC. The result is premium quality siRNA that is purified and ready to use.
Technical inquires:
Our Technical Application Scientists are available to help assist you at techsupport@thermofisher.com
Ordering & Order Status inquires:
If you have questions about pre-designed RNAi orders and order status, please contact us at genomicorders@thermofisher.com
If you have any questions about Custom RNAi orders and order status, please contact us at RNAiSupport@thermofisher.com
For Research Use Only. Not for use in diagnostic procedures.