Search
The MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit (Cat. No. A48383) is now compliant with In Vitro Diagnostic Regulation (EU) 2017/746. This kit is designed to recover viral nucleic acids from nasopharyngeal swabs and viral RNA from saliva specimens. The kit utilizes MagMAX magnetic-bead technology, enabling reproducible recovery of high-quality nucleic acid and is compatible with the KingFisher Flex instrument (GPLE).
For information on the MagMAX Viral/Pathogen II Nucleic Acid Isolation kit for research use, please visit the MVP II (RUO)page.
For current information on the Sample Prep and Molecular Biology IVDR transition, please visit the Sample Prep and Molecular Biology IVDR Transition Updates page.
![]() MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit Order now | |
| Intended use | The MagMAX Viral/Pathogen II Nucleic Acid Isolation Kit (Cat. No. A48383) is a nucleic acid purification kit based on magnetic bead technology. The kit is intended for the isolation and purification of viral nucleic acids from human nasopharyngeal swabs and viral RNA from saliva specimens, and is suitable for use in molecular detection by qPCR. The kit is intended for use by qualified and trained clinical laboratory personnel specifically instructed and trained in the techniques of magnetic bead purification, either manual or automated, and in vitro diagnostic procedures. |
| Sample types | Human nasopharyngeal swabs and saliva |
| Automation level | Automation-ready protocols compatible with Thermo Scientific KingFisher Flex purification system |
| Sample input volume | 200 or 400 µL of VTM |
| Elution volume | 50 μL |
| Time per run | 96 samples can be processed in <30 minutes |
| Shelf life | 24 months |
| Assay compatibility | Multiple downstream assays have confirmed MVP II as compatible. Visit the assays IFU for more information. |
| Kit Configuration | Now available as a convenient single-box format with a single expiration date. |
| Cat. No. | A48383 (up to 2,000 preps/kit) |
The 24 shelf life extension applies in select countries only and availability is subject to terms and conditions. Please check with your country specific representative for more information.
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.