Scale confidently with customizable, high-potency siRNA libraries. High-throughput screens harness the power of RNAi to better understand gene function and biological pathways. Thermo Fisher Scientific offers predefined and custom sets of potent siRNAs for arrayed functional genomics studies. Each of our siRNA libraries is backed by a silencing guarantee, helping give you confidence in the hits you identify and supporting reliable results.
We offer Silencer Select and Silencer siRNA libraries, as either predefined or custom libraries. All our libraries are included with full siRNA annotation files to include:
- Plate map ID and well address for each siRNA
- siRNA sequence information
- NCBI identifiers: gene symbol, gene name, alias(es), Entrez gene ID, and associated RefSeq mRNA accession numbers.
- Plates are individually barcoded with unique identifiers
Silencer Select siRNA libraries
Silencer Select siRNA is our most potent and specific siRNA available due to a next-generation algorithm and patented chemical modifications.
Available Silencer Select siRNA library options include:
- Human pre-defined gene families and pathways, including the druggable genome
- Human whole genome siRNA library
- Human genome-wide siRNA collection to target long-noncoding genes
Advantages of Silencer Select siRNA libraries
Backed by one of the best silencing guarantees in the industry, our Silencer Select siRNA libraries are designed to be reliable, efficient and specific for clear and consistent siRNA screening results.
| Attributes | Advantages |
|---|
- Advanced bioinformatic filters for sequence selection
- LNA chemical modifications
| - Reduced off-target effects by up to 90% for fewer false positives
- Less toxicity due to off-target effects
- More consistent phenotypic data
|
- Next-generation design algorithm
| - Up to 100-fold more potent silencing than other available siRNAs
- Improved silencing for more robust knockdown results and consistent results
- Fewer false negatives for greater experimental confidence
|
- Three unique individual siRNAs per gene
| - Multiple data points per gene for rapid progression from primary screening to hit confirmation studies
|
Silencer siRNA libraries
Silencer siRNA is a great choice for cost-effective screening of pre-defined gene family subsets and popular pathways to accelerate your functional genomics studies. These unmodified siRNA screening libraries are built with our Silencer predesign inventory and achieve good silencing at a low cost.
Predefined Silencer Select siRNA libraries
These libraries have been pre-assembled using PANTHER and Gene Ontology™ annotation information to create up-to-date target gene sets.
Ordering siRNA libraries
- Each gene is targeted by three unique, non-overlapping siRNAs.
- Validated siRNAs are included at no extra cost.
- 0.25 nmol of each siRNA is included dried-down in the wells of barcoded 96- or 384-well plates. This is sufficient for 100–500+ transfections, depending on siRNA working concentration and reaction volume.
- Column 12 of each 96-well plate and columns 23 and 24 in each 384-well plate are left empty to allow for the addition of controls at the time of use.
- Plates are conveniently arranged for pooling of siRNA, if desired.
If you would like more information on any of the libraries listed below, click the product link or email us at RNAiSupport@thermofisher.com.
Ordering information—Silencer Select siRNA libraries
A30085, A30087, A30089, A30091, A30092, A30095, A30133, A30136, A30138, A30140, A30142, A30144, A30146, A30148, A30149, 4397914, 4397915, 4397916, 4397917, 4397918, 4397919, 4397926, 4397923, 4397920, 4397921, 4397927
Predefined Silencer siRNA libraries
These libraries have been pre-assembled using PANTHER and Gene Ontology™ annotation information to create up-to-date target gene sets.
- Each gene is targeted by three unique siRNAs.
- Validated siRNAs are included at no extra cost.
- Plates are conveniently arranged for pooling of siRNA, if desired.
- 0.25 nmol of each siRNA is included dried-down in the wells of barcoded 96-well plates.
- Column 12 of each 96-well plate is left empty to allow for the addition of controls at the time of use.
If you would like more information on any of the libraries listed below, click the product link or email us at RNAiSupport@thermofisher.com.
Ordering information- Silencer siRNA libraries
A30079, A30080, A30081, A30082, A30083, A30084, A30086, A30088, A30090, A30096, A30135, A30137, A30139, A30141, A30143, A30145, A30147
Custom siRNA libraries
Custom Silencer Select and Silencer siRNA libraries are available to suit your specific research needs. These highly economical plated versions of our guaranteed siRNAs are an excellent choice for pilot studies, primary screen follow-up, or a customized functional genomics screen. Plate layouts and gene lists can be customized based on your experimental requirements.
- Minimum order of 20 wells, which can include positive and negative controls
- Pooled siRNA options available—Choose from 96-well, 384-well, or 384-well Echo-qualified plates
- Validated siRNAs are available at no additional cost
| Product Line for custom siRNA libraries | Quantity per well* |
|---|
| Silencer Select human predesigned siRNA | 0.1, 0.25, 0.5, 1, or 2 nmol |
| Silencer human predesigned siRNA | 1, 2, or 5 nmol |
| *NOTE: Not all quantities available in 384-well |
Predefined Silencer Select siRNA libraries
These libraries have been pre-assembled using PANTHER and Gene Ontology™ annotation information to create up-to-date target gene sets.
Ordering siRNA libraries
- Each gene is targeted by three unique, non-overlapping siRNAs.
- Validated siRNAs are included at no extra cost.
- 0.25 nmol of each siRNA is included dried-down in the wells of barcoded 96- or 384-well plates. This is sufficient for 100–500+ transfections, depending on siRNA working concentration and reaction volume.
- Column 12 of each 96-well plate and columns 23 and 24 in each 384-well plate are left empty to allow for the addition of controls at the time of use.
- Plates are conveniently arranged for pooling of siRNA, if desired.
If you would like more information on any of the libraries listed below, click the product link or email us at RNAiSupport@thermofisher.com.
Ordering information—Silencer Select siRNA libraries
A30085, A30087, A30089, A30091, A30092, A30095, A30133, A30136, A30138, A30140, A30142, A30144, A30146, A30148, A30149, 4397914, 4397915, 4397916, 4397917, 4397918, 4397919, 4397926, 4397923, 4397920, 4397921, 4397927
Predefined Silencer siRNA libraries
These libraries have been pre-assembled using PANTHER and Gene Ontology™ annotation information to create up-to-date target gene sets.
- Each gene is targeted by three unique siRNAs.
- Validated siRNAs are included at no extra cost.
- Plates are conveniently arranged for pooling of siRNA, if desired.
- 0.25 nmol of each siRNA is included dried-down in the wells of barcoded 96-well plates.
- Column 12 of each 96-well plate is left empty to allow for the addition of controls at the time of use.
If you would like more information on any of the libraries listed below, click the product link or email us at RNAiSupport@thermofisher.com.
Ordering information- Silencer siRNA libraries
A30079, A30080, A30081, A30082, A30083, A30084, A30086, A30088, A30090, A30096, A30135, A30137, A30139, A30141, A30143, A30145, A30147
Custom siRNA libraries
Custom Silencer Select and Silencer siRNA libraries are available to suit your specific research needs. These highly economical plated versions of our guaranteed siRNAs are an excellent choice for pilot studies, primary screen follow-up, or a customized functional genomics screen. Plate layouts and gene lists can be customized based on your experimental requirements.
- Minimum order of 20 wells, which can include positive and negative controls
- Pooled siRNA options available—Choose from 96-well, 384-well, or 384-well Echo-qualified plates
- Validated siRNAs are available at no additional cost
| Product Line for custom siRNA libraries | Quantity per well* |
|---|
| Silencer Select human predesigned siRNA | 0.1, 0.25, 0.5, 1, or 2 nmol |
| Silencer human predesigned siRNA | 1, 2, or 5 nmol |
| *NOTE: Not all quantities available in 384-well |
Publications featuring Silencer Select siRNA libraries
Since 2010, Silencer Select siRNA libraries have been enabling functional genomics studies in labs around the world. These selected peer-reviewed references showcase the power of our high-quality siRNAs in high-throughput screening applications.
- Hasson S, Kane L, et al. (2013) High-content genome-wide RNAi screens identify regulators of parkin upstream of mitophagy. Nature 504: 291-295.
This Nature publication demonstrates the utility of the Silencer Select genome-wide siRNA library for arrayed high-throughput screening in effectively identifying novel drug targets for Parkinson’s disease. - Shum D, Bhinder B, et al. (2013) An arrayed RNA interference genome-wide screen identifies candidate genes involved in the MicroRNA 21 biogenesis pathway. Assay Drug Dev Technol 11: 191-205.
Researchers at Memorial Sloan Kettering uncover a number of novel candidate regulators of microRNA function using the Silencer Select whole genome siRNA library. - Sivan G, Ormanoglu P, et al. (2015) Identification of Restriction Factors by Human Genome-Wide RNA Interference Screening of Viral Host Range Mutants Exemplified by Discovery of SAMD9 and WDR6 as Inhibitors of the Vaccinia Virus K1L−C7L− Mutant. mBio 6: e01122-15.
Genome-wide screen results demonstrate higher phenotypic penetrance from multiple individual Silencer Select siRNAs than with a pooled siRNA reagent from another vendor. - Lingeman M, McCarty T, et al. (2019) The alpha-1 subunit of the Na+,K+-ATPase (ATP1A1) is required for macropinocytic entry of respiratory syncytial virus (RSV) in human respiratory epithelial cells. PLoS Pathog 15: e1007963.
A genome-wide Silencer Select siRNA library screen uncovers a model for RSV entry into human airway epithelial cells requiring activation of ATP1A1, signaling it as a potential new target for the development of anti-RSV drugs. - Kainulainen, V., von Schantz-Fant, C., et al. (2022) Genome-wide siRNA screening reveals several host receptors for the binding of human gut commensal Bifidobacterium bifidum. npj Biofilms Microbiomes 8, 50
Genome-wide Silencer Select screen reveals cell surface proteins involved in B. bifidium-host cell interactions. - Shrestha , R., Balachandra, V., et al. (2023) The histone H3/H4 chaperone CHAF1B prevents the mislocalization of CENP-A for chromosomal stability. J Cell Sci 136: 10
The NIH defines gene roles in preventing CENP-A mislocalization and chromosomal instability using an imaging-based high-throughput siRNA screen with a Silencer Select Epigenetics library. - Morimoto, K., Yamada, T., et al. (2024) AXL signal mediates adaptive resistance to KRAS G12C inhibitors in KRAS G12C-mutant tumor cells. Cancer Letters 587: 216692
Applying the Silencer Select Kinase library, researchers investigate the mechanism for adaptive resistance to KRAS G12C inhibitors for treatment in lung cancer. - Kolapalli, S., Beese , C., et al. (2025) Pellino 3 E3 ligase promotes starvation-induced autophagy to prevent hepatic steatosis. Sci Adv 11, eadr2450
Researchers use the Silencer Select Ubiquitin library to identify crucial genes involved in regulating starvation-induced autophagy with high-content imaging.