Explore a broad array of microplate assays by modality, application, and instrument compatibility.
Microplate assays
Category
Product
Wavelength
Multiskan FC Multiskan SkyHigh Varioskan ALF
Varioskan LUX
Cell viability alamarBlue Cell Viability Reagent  570 nm
Cell viability alamarBlue HS Cell Viability Reagent 570 nm
Cell viability CyQUANT LDH Cytotoxicity  590 nm
Cell viability CyQUANT MTT Cell viability Assay 570 nm
Cell viability CyQUANT XTT Cell viability Assay 450 nm and 660 nm
Cell viability PrestoBlue Cell viability Reagent 570 nm
Cell viability PrestoBlue HS Cell viability Reagent 570 nm
ELISA Invitrogen ELISA Kits (Coated or Uncoated) - Choose from >2500 targets 450 nm
Endotoxin quantification Pierce Chromogenic Endotoxin Quant Kit 405 nm
Peptide Assays Pierce Dilution-Free Rapid Gold BCA Protein Assay 480 nm
Protein assays Pierce BCA Protein Assay Kit with Dilution-Free BSA Protein Standards 562 nm
Protein assays Pierce Bradford Plus Protein Assay Kit with Dilution-Free BSA Protein Standards 595 nm
Protein assays Micro BCA Protein Assay Kit  562 nm
Protein assays Pierce Microplate BCA Protein Assay Kit - Reducing Agent Compatible 562 nm
Protein assays Pierce BCA Protein Assay Kit - Reducing Agent Compatible  562 nm
Protein assays Pierce Detergent Compatible Bradford Assay Kit 595 nm
Protein assays Pierce 660nm Protein Assay Kit 660 nm
Protein assays Pierce Modified Lowry Protein Assay Kit 750 nm
Protein assays Pierce Quantitative Colorimetric Peptide Assay 480 nm
Category
Product
Wavelength
Multiskan FC Multiskan SkyHigh Varioskan ALF Varioskan LUX
Cell function reagents ATP Determination Kit 560 nm    
Cell viability QuantiGene Singleplex Gene Expression Assay Kit      
Cell viability Luc-Screen Extended-Glow Luciferase Reporter Gene Assay System      
Cell viability NovaBright Phospha-Light EXP Assay Kit for SEAP Reporter Gene Detection       
Cell viability Galacto-Star β-Galactosidase Reporter Gene Assay System for Mammalian Cells      
Category
Product
Wavelength
Multiskan FC Multiskan SkyHigh Varioskan ALF Varioskan LUX
Peroxidase assay Amplex Red Hydrogen Peroxide/Peroxidase Assay Kit 571/585 nm    
Cell proliferation Click-iT EdU Proliferation Assay for Microplates 568/585 nm      
Cell proliferation CyQUANT Cell Proliferation 480/520 nm      
Cell proliferation CyQUANT NF (No Freeze) Cell Proliferation 497/520 nm      
Cell signaling and lipids Amplex Red Cholesterol Assay Kit 571/585 nm    
Cell signaling and lipids EnzChek Phosphatase Assay Kit 360/460 nm    
Cell viability alamarBlue Cell viability Reagent 560/590 nm    
Cell viability alamarBlue HS Cell viability Reagent 560/590 nm    
Cell viability CyQUANT Cytotoxicity Assay Kit (G6PD Release Assay) 560/590 nm    
Cell viability CyQUANT LDH Cytotoxicity Assay, fluorescence 560/590 nm    
Cell viability LIVE/DEAD BacLight Bacterial Viability Kit 485/530 nm
630 nm
Cell viability LIVE/DEAD Viability/Cytotoxicity Kit, for mammalian cells 494/517 nm 517/617 nm      
Cell viability LIVE/DEAD Yeast Viability Kit 485/530 nm      
Cell viability PrestoBlue Cell viability Reagent 560/590 nm    
Cell viability PrestoBlue HS Cell viability Reagent 560/590 nm    
Endotoxin quantification Quant-iT Endotoxin Detection Assay Kit 490/525 nm    
Enzyme activity EnzChek Protease Assay Kit, green fluorescence  505/513 nm    
Enzyme activity EnzChek Protease Assay Kit, red fluorescence 589/617 nm    
pH indicators pHrodo Green AM Intracellular pH indicator 509/533 nm    
pH indicators pHrodo Red AM Intracellular pH indicator 560/585 nm    
Ion indicators Measure-IT Thiol Assay Kit      
Ion indicators FluoVolt Membrane Potential Kit      
Ion indicators Fluo-4 Direct Calcium Assay Kit      
Ion indicators Fura-2, AM, cell permeant      
Ion indicators FluxOR II Green Potassium Ion Channel Assay      
Nucleic acids Quant-IT dsDNA Assay Kit, high sensitivity (HS) and broad range (BR) 502/523 nm    
Nucleic acids Quant-iT 1X dsDNA Assay Kit, high sensitivity (HS) and broad range (BR) 502/523 nm    
Nucleic acids Quant-iT PicoGreen dsDNA Assay Kits and Reagents 502/523 nm    
Nucleic acids Quant-iT PicoGreen ReadyPlate 502/523 nm    
Nucleic acids Quant-iT microRNA Assay Kit 500/525 nm    
Nucleic acids Quant-iT OliGreen ssDNA Assay Kit and Quant-iT OliGreen ssDNA Reagent 500/525 nm    
Nucleic acids Quant-iT RiboGreen RNA Assay Kit and Reagent 500/525 nm    
Nucleic acids Quant-iT RiboGreen ReadyPlate 500/525 nm    
Nucleic acids Quant-iT RNA Assay Kit and Quant-iT RNA HS Reagent  644/673 nm    
Peptide assays Pierce Quantitative Fluorometric Peptide Assay 390/475 nm    
Protein assays CBQCA Plus Protein Quantitation Kit 465/550 nm    
Protein assays NanoOrange Protein Quantitation Kit 485/590 nm    
Protein assays Quant-iT Protein Assay Kit 485/590 nm    
Protein assays EZQ Protein Quantitation Kit 485/590 nm    
Reactive oxygen species CellROX Deep Red Reagent, for oxidative stress detection 644/665 nm    
Reactive oxygen species CellROX Green Reagent, for oxidative stress detection 485/520 nm    
Category
Product
Wavelength
Multiskan FC Multiskan SkyHigh Varioskan ALF Varioskan LUX
  LanthaScreen Cellular Assays         
Virtual demo: Simplify Fluorescence Assays