Search
For a general-purpose mass spectrometer look no further than the Thermo Scientific Linear Ion Trap Mass Spectrometer. With new lower prices, Ion trap mass spectrometers are cost-effective and provide automation features which enable all users, despite expertise level, to be more confident in not only their results, but instrument operation skills. Learn more about ion trap mass spectrometry below or request assistance from one of our experts.
As the main RF voltage increases, ions of greater and greater mass-to-charge ratios eject through the slots in the linear trap. Ions are focused toward the ion detection system where they are detected.
Listen directly from our expert, Julie Horner, about the benefits of Ion Trap mass spectrometry for various applications.
| Performance Specifications | For everyday affordability![]() LTQ XL Linear Ion Trap Mass Spectrometer | |||
| Description | Cost effective and flexible. | |||
| Sensitivity | 250 fg Reserpine; 100:1 S/N | |||
![]() | ||||
| Linear Dynamic Range | >4.5 orders of magnitude | |||
![]() | ||||
| Mass Ranges |
| |||
![]() | ||||
| Resolution, scan speed and mass accuracy | Peak Width (FWHM) | Scan Rate (Da/Sec) | Mass Accuracy (Da) | |
| 3 | 125,000 | 1.5 | ||
| 0.7 | 16,667 | 0.15 | ||
| 0.45 | 5,000 | 0.15 | ||
| 0.3 | 1,111 | 0.15 | ||
| 0.15 | 28 | 0.15 | ||
![]() | ||||
| Application across multiple matrices | ||||
| Proteomics Peptide Identification | Ion Trap![]() | Q Exactive Plus![]() | ||
| Pharmaceutical Metabolism and Metabolite Identification | Ion Trap![]() | Q Exactive Plus![]() | ||
| Untargeted Screening | — | Ion Trap![]() | ||
| Unknown Screening | Ion Trap![]() | Q Exactive Plus![]() | ||
| Structural Elucidation and Confirmation | Q Exactive Plus![]() | Ion Trap![]() | ||
| Quantitation | Q Exactive Plus![]() | |||
= Good | = Better | = Best |
| Structural Characterization | LTQ XL |
|---|---|
Collision Induced Dissociation (CID)
| Included |
Pulsed “q” Dissociation (PQD)
| Included |
Higher Energy Collision Induced Dissociation (HCD)
| Not Available |
Achieve high quality full-scan and MS/MS analysis of samples quickly, and easily for running your application or for teaching students the fundamentals of mass spectrometry. Take control of your data using the efficient Thermo Scientific Xcalibur software which enables method set up, acquisition, processing and reporting.
Take full advantage of new lower prices offered by the highest capacity, most flexible ion trap MS instruments on the market. We
offer customized service offerings with Unity Lab Services designed to meet your service support needs.
PaperSpray technology is a direct ionization technique for analysis of small molecules from complex matrices such as blood or urine. Enables quick, sensitive and selective analysis from sample to mass spectrometer.
Our LC-MS on-demand webinar series expands your application knowledge for pharma, biopharma, environmental, omics, food, forensics and clinical research and more. Learn how the latest high-performance mass spectrometry solutions can enable your laboratory to solve your most pressing analytical challenges, regardless of sample type.