Search
POROS non-affinity chromatography columns are designed for reverse phase, anion exchange and cation exchange chromatography of peptides, proteins, polynucleotides and other biomolecules in the high-resolution separation mode. They consist of polymeric packing of cross-linked poly[styrene-divinylbenzene] flow-through particles for rapid mass transport.
Check out the new Chromatography Learning Lab from Thermo Fisher Scientific, providing a robust range of webinars, articles, eBooks, infographics, and other digital resources to help you optimize the use of our purification products in your process.
Explore digital resource
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing: