Introduction
There is great interest in immuno-oncology research as more biotherapeutics have shown success in clinical trials. These new drugs take many forms, including chimeric antigen receptor (CAR) T cells, checkpoint blockade antibodies, and T cell–engaging bispecific antibodies. All of these immunotherapy strategies improve the ability of T lymphocytes to target and kill cancer cells.
Research and development of new immunotherapies requires robust techniques to purify and culture T cells. Here we describe methods for isolation of T cells from blood followed by activation and expansion using Invitrogen Dynabeads magnetic beads without the need for feeder cells or antigens. Overstimulation of T cells can lead to activation-induced cell death, and variations in cell culture conditions or technique can affect T cell viability.
Obtaining maximum viability is critical to the optimal performance of T cells in functional assays. It is important to be able to quickly assess the health of T cells using a small sample of cells. The Invitrogen Countess II FL Automated Cell Counter provides fast, accurate measurements of cell viability and concentration that can be performed both before and after isolation and activation, saving time and precious cells for future experiments. Additionally, the Invitrogen EVOS XL Core Imaging System can be used to monitor the expansion of T cells without disturbing the activation process or requiring extended time outside of the incubator.
Methods
Isolation of T cells from whole blood
Human peripheral blood was mixed with an equal volume of PBS (20 mL each) and carefully layered over 15 mL of Ficoll solution and centrifuged at 400 x g for 30 minutes with minimal acceleration and deceleration. The peripheral blood mononuclear cell (PBMC) layer was isolated and washed with PBS. The cells were mixed with an equal volume of trypan blue stain (10 μL each), and 10 μL of the mixture was pipetted into the chambers of a Countess slide (Figure 1). The Countess II FL Automated Cell Counter was used to determine cell viability and concentration. Cell concentration was then adjusted with PBS to 1 x 108 cells/mL.
Countess chamber slide with one chamber filled and the second chamber in mid-fill with pipette tip
Figure 1. Loading a Countess chamber slide.
Negative isolation of T cells was performed using the Dynabeads Untouched Human T Cells Kit as described in the product insert. Briefly, the provided antibody mixture and Depletion Dynabeads magnetic beads were used to remove B cells, natural killer (NK) cells, monocytes, platelets, dendritic cells, granulocytes, and erythrocytes, leaving T cells for further use.
Activation of human T cells
Isolated T cells were analyzed on the Countess II FL Automated Cell Counter to determine cell viability and concentration. Cell concentration was adjusted to 1 x 106 cells/mL in CTS OpTmizer T Cell Expansion SFM. Dynabeads Human T-Activator CD3/CD28 was added at a bead:cell ratio of 1:1. Cells were plated in a T-75 flask in a cell culture incubator at 37°C with 5% CO2.
Monitoring of T cell proliferation and viability
Every 24 hours, the flask was removed from the incubator and viewed at 40x magnification on an EVOS XL Core Imaging System. Initially the T cells and beads were observed to settle to the bottom of the flask. After 24 hours, the cells and beads began to bind and form clumps that grew in size as the cells proliferated. The cell clumps were weakly attached and could easily be disrupted by pipetting the medium up and down after 5 days of expansion. Using normal cell counting techniques it may be difficult to obtain an accurate measure of cell concentration when the beads are still in the culture flask. The Countess software allows cells to be gated based on size, brightness, and circularity, allowing easy inclusion or exclusion of debris or specific cell populations. Increasing the lower size threshold sets the instrument to only count the larger T cells and not the smaller beads.
Once the desired expansion was completed, the beads and cells were resuspended in medium and transferred to a 15 mL conical tube. The tube was then placed in a DynaMag-15 Magnet for 2 minutes. The beads bound to the side of the tube while purified, activated T cells were removed along with the medium and could be used for future experiments.
Results
The Dynabeads Untouched Human T Cells Kit was used to prepare isolated T cells that were 95% viable as determined by the Countess II FL Automated Cell Counter (Figure 2).
These isolated T cells were then activated and expanded with Dynabeads Human T-Activator CD3/CD28. This process could be easily monitored by viewing the flask of T cells and beads on the EVOS XL Core Imaging System without disturbing the cells or the activation process (Figures 3 and 4).
Screen capture from Countess cell counter showing total concentration and percentage of live and dead cells
 Click image to enlarge
Figure 2. Viability and concentration of isolated T cells determined using the Countess II FL Automated Cell Counter. T cells isolated with the Dynabeads Untouched Human T Cells Kit were 95% viable.
Viability and concentration of the activated T cells were measured on the Countess II FL Automated Cell Counter. Beads were excluded from the cell count by increasing the size threshold using the gating feature of the instrument software. Viability of the activated T cells could be determined even in the presence of beads (Figure 5).
Conclusion
This workflow enables the isolation, activation, and expansion of robust T cells for downstream immunooncology applications, a critical need in this research area for cancer treatment and therapeutics.
The Countess II FL Automated Cell Counter provides fast, accurate measurements of T cell viability and concentration both after initial isolation and activation, even when Dynabeads Human T-Activator CD3/CD28 is present. These critical in-process measurements help determine if the cells are healthy and should continue to be expanded. Dynabeads Human T-Activator CD3/CD28 provides strong activation of T cells grown in CTS OpTmizer T Cell Expansion SFM. The EVOS XL Core Imaging System enables a quick check of T cell proliferation (indicated by cell clumping) without disruption of the cells or extended time outside the incubator. The combination of these reagents with the ability to monitor T cell activation, proliferation, and viability with minimal cell disturbance enables production of high-quality T cells that can be used in immuno-oncology experiments to help accelerate knowledge in this important area of cancer therapeutics.
Ficoll is a trademark of GE Heathcare.
For Research Use Only. Not for use in diagnostic procedures.