Search
The ability to assess cell health in a cell population is a basic and critical evaluation parameter in many cell and molecular biology labs. To this end, knowing how many cells are dead or dying are key pieces of information. To answer such basic questions, the cells of interest can be stained with a viability dye, such as Molecular Probes™ SYTOX™ Red Dead Cell Stain, as well as an apoptosis indicator that measures caspase activation, such as Molecular Probes™ CellEvent™ Caspase-3/7 Green Detection Reagent. This staining combination, used together with an Invitrogen™ Countess™ II FL Automated Cell Counter equipped with GFP and Cy®5 EVOS™ Light Cubes, allows a quick and simple method to obtain apoptosis data together with cell viability data.
The CellEvent Caspase-3/7 Green Detection Reagent (Ex/Em: 502/530 nm when bound to DNA) is intrinsically nonfluorescent, as the DEVD peptide inhibits the ability of the dye to bind to DNA. However, after activation of caspase-3/7 in apoptotic cells, the DEVD peptide is cleaved, enabling the dye to bind to DNA and produce a bright, fluorogenic response. The fluorescence emission of the dye when bound to DNA can be observed using a standard “FITC” or GFP filter set.
To use CellEvent Caspase 3/7 Green Detection Reagent, simply add substrate to control and treated cells, incubate 30 minutes, and visualize. Apoptotic cells with activated caspase-3/7 will have bright green nuclei, while cells without activated caspase 3/7 will have minimal fluorescence signal.
SYTOX dead cell stains do not cross intact cell membranes, and they exhibit increased fluorescence upon dsDNA binding, making them some of our brightest dead cell stains. SYTOX dead cell stains can be applied to cells and visualized without an additional wash step because they are nonfluorescent in aqueous media. These stains are available in multiple single-color formats, making them compatible with many filter sets.
After brief incubation with SYTOX Red Dead Cell Stain, the nucleic acids of dead cells fluoresce bright red when excited and detected with traditional Cy5 filter sets. SYTOX Red Dead Cell Stain is distinct from dead cell probes such as 7-AAD and propidium iodide (PI) that are excited using 488 nm light. Moreover, the emission of SYTOX Red Dead Cell Stain is limited to one channel, thus minimizing spectral overlap with CellEvent Caspase 3/7 Green Detection Reagent and simplifying workflow.
What used to take more than an hour on traditional microscopes or cytometers—simply acquiring preliminary results—now takes only a few minutes with the Countess II FL Automated Cell Counter. You can save time and effort by checking your sample before using a microscope or flow cytometer.
Figure 1. Apoptotic and dead cells counted on a Countess II FL Automated Cell Counter. A Countess II FL Automated Cell Counter was loaded with EVOS Light Cube for GFP and EVOS Light Cube for Cy5. After incubation with 0.5 µm staurosporine, HeLa cells were labeled with 1:400 CellEvent Caspase-3/7 Green Detection Reagent to identify apoptotic cells, and then stained with 1:1,000 SYTOX Red Dead Cell Stain to denote all dead cells and incubated at room temperature for 30 minutes.
Figure 2. Apoptosis of HeLa cells measured on a Countess II FL Automated Cell Counter using CellEvent Caspase-3/7 Green Detection Reagent and SYTOX Red Dead Cell Stain. The indicated amounts of staurosporine were applied to HeLa cells to trigger apoptosis. Increases in green and far-red signal were noted with increased drug treatment, indicating significant increases in apoptosis and cell death, respectively.
For Research Use Only. Not for use in diagnostic procedures.