Search
Invitrogen EVOS imaging systems are designed to help eliminate the complexities of high-end microscopy without compromising performance. Remarkably versatile and intuitive, EVOS microscopes are excellent for a broad range of imaging applications, including those involving both cell culture and tissue samples.
The EVOS M3000 Imaging System is a manual microscope capable of up to 2-color fluorescence, designed to be the workhorse of any cell culture lab with the ability to report confluency percentage seen in the field of view in less than one second. The EVOS M5000 Imaging System is a semi-automated microscope capable of up to 4-color fluorescence that can be configured to excel in both cell culture and imaging, providing a flexible solution for diverse research needs. The EVOS M7000 Imaging System is a fully automated microscope capable of up to 4-color fluorescence with advanced 2D deconvolution capabilities designed to simplify the most demanding 2D and 3D tissue and cell imaging applications. The EVOS S1000 Spatial Imaging System streamlines the process of tissue imaging for spatial localization of proteins.
All EVOS systems deliver publication-quality images and data in no time, over time, every time at an exceptional value.
For Research Use Only. Not for use in diagnostic procedures.