On this page:
PrepSEQ chemistry-based sample preparation and automation
PrepSEQ chemistry is a universal sample preparation workflow for contaminant and impurity testing used in multiple applications, including residual host cell DNA quantitation, mycoplasma detection, Minute Mouse Virus (MMV), and viral detection.
Key system features:
  • Easy-to-use protocol featuring magnetic bead-based technology for efficient sample extraction, clean up, and elution
  • Sensitive isolation of genomic DNA from total lysate of test samples
  • Increased throughput capabilities with flexible automation formats
  • Rapid routine screening of high-value cell lines in combination with the resDNASEQ Quantitative Residual Host Cell DNA Kit
  • Accurate determination of absence or presence of contaminants with the MycoSEQ Mycoplasma Detection Kit
Universal sample preparation for impurity and contaminant testing
Get reliable, consistent quantitative recovery of your target DNA and RNA with a comprehensive solution that is built on years of successful performance. PrepSEQ technology enables consistent recovery from matrices varying in complexity, whether it is pH, salt quantity, or protein balance. The nucleic acid extraction workflow includes our proprietary magnetic particle and binding solution that not only enables high binding but also complete elution of the purified sample. The PrepSEQ solution offers flexible protocols for manual sample preparation or automation protocols, allowing you to process 1–96 extractions at one time, with high recovery to help enable you to meet regulatory guidelines.
PrepSEQ nucleic acid extraction workflow
Flexible sample preparation solutions for qPCR
PrepSEQ solutions are well suited for a wide range of applications in the biopharma industry. These applications include residual host cell DNA quantitation, residual plasmid DNA quantitation, mycoplasma detection, and viral detection. These comprehensive solutions enable you to address all your analytical needs with confidence and precision.
Our technology helps ensure the recovery of all forms of nucleic acid, including double-stranded DNA, single-stranded DNA, and RNA. This complete nucleic acid recovery capability allows for accurate and efficient analysis, supporting a variety of testing and research applications.
Automated sample preparation solutions
Help boost lab productivity by using our automated solutions designed to reduce cost and hands-on sample preparation time, increase throughput, and improve the quality of sample extractions. Intuitive to operate and pre-loaded with protocols, our automated sample preparation instruments help meet your throughput needs. Our automated solutions offer multi-assay, timesaving, cost-effective DNA extraction for residual host cell DNA, mycoplasma, MMV, and Vesivirus.
Sample preparation solutions for residual DNA and RNA quantitation
PrepSEQ Residual DNA Sample Preparation Kits are optimized for consistent recovery of DNA and RNA from various sample matrices. The kit has flexible protocols for both manual and automated workflows to enable consistent, reliable recovery for even the most challenging samples. The PrepSEQ Residual DNA Sample Prep Kit can be used with the resDNASEQ quantitative residual host cell DNA kits that are available for CHO, Human, HEK293, E. coli, Vero, MDCK, NS0, and Pichia pastoris cell lines.
We offer a high throughput solution for the automation of the PrepSEQ workflow to purify samples for residual host cell DNA analysis. The complete sample DNA and RNA extraction and 96-well preparation for qPCR analysis can be achieved in as little as three hours using PrepSEQ/resDNASEQ kits. The KingFisher Apex and KingFisher Flex Instruments automate the PrepSEQ workflow to help deliver reliable high DNA/RNA recovery with consistent performance, regardless of sample complexity.
The PrepSEQ Nucleic Acid Sample Preparation Kit is optimized for highly efficient, quantitative, and consistent recovery of DNA and RNA from diverse samples types, including cell culture, raw material (e.g., serum), and in-process downstream purification samples, such as column purification or bulk drug substance samples.
Sample preparation solutions for contaminant detection
Conduct manual sample preparation for the extraction of DNA and RNA from mycoplasma cells, MMV isolates, and/or Vesivirus isolates. Minimizing cellular DNA and/or RNA in the final sample extraction is critical to mycoplasma DNA detection where high amounts of cellular DNA and RNA can cause PCR inhibition and high background. PrepSEQ 1-2-3 kits allow for sample cleanup of mycoplasma genomic DNA from research cell lines, stem cell lines, cellular, and tissue therapy samples with high efficiency. The workflow offers a rapid and easy to use protocol for results as little as 2 hours.
We offer an easy-to-use benchtop instrument that enables hands-free automation of the nucleic acid extraction process for the MycoSEQ Mycoplasma Detection, ViralSEQ MMV Detection, and ViralSEQ Vesivirus Detection testing workflows. The instrument uses the established PrepSEQ nucleic acid extraction chemistry packaged into a cartridge format for quantitative recovery of DNA and RNA. The Automate Express System offers out-of-the-box automation for easy integration into existing workflows and lab configurations. There is minimal hands-on time with this closed system, allowing you to “load and go” with prefilled reagent cartridges and preprogrammed protocols. The Automate Express magnetic particle processor instrument offers low to medium throughput of up to 13 extractions in one run. 
This ready-to-go kit contains pre-filled reagents and plastic tubes and tips to perform hands-free extraction of purified nucleic acid from 52 samples. The kit is designed for use on the AutoMate Express Nucleic Acid Extraction System. The cartridges utilize the established PrepSEQ nucleic acid extraction chemistry for quantitative recovery of DNA and RNA from diverse samples types, including cell culture, raw material, and in-process samples.
Analytics Knowledge Hub
Explore the Analytics Knowledge Hub and discover a robust collection of webinars, articles, eBooks, infographics, and other digital resources to help you optimize analytical testing in your bioprocess workflow.
Request a technical consultation
No matter your questions, we are here to help. Our experienced team of scientists and technical specialists is ready to assist you in choosing the right solution and products. Complete this form to start the conversation.
Additional bioprocessing resources
Discover areas where you can cut costs and gain efficiency using our integrated calculator.
Explore our collection of information and insights to support your drug development journey.
Learn, evaluate, connect.
Connect with a Bioprocessing representative or request more info.
For Research Use Only. Not for use in diagnostic procedures.