A broad array of channel options and confocal capabilities
Our HCS/HCA family of platforms includes options for laser or LED fluorescence illumination, widefield and brightfield modes, and confocal capabilities. Compare platforms to find the one that fits your research.
Check out the new CX7 Pro series
Which high-content screening platform is right for you?
All of our high-content screening platforms offer high-speed image acquisition, powerful image processing algorithms and unparalleled real-time analysis. Platforms feature more than 30 validated assays and options including live-cell imaging and in-depth life sciences analysis, including single-cell analysis capabilities. Choosing the platform that is right for you depends on your needs and applications.
Our high-content screening platforms support the full range of imaging needs including broad array of channel options and confocal capabilities. Browse the categories and specifications below, then link to your models of choice for more information.
Selection guide
CellInsight CX7 LZR Pro
CellInsight CX7 Pro CellInsight CX5
Superior High-Content Screening performance for lightning-fast discovery.  Incorporates both laser-based fluorescence illumination and Nipkow spinning-disc confocal technology to illuminate a broad range of samples from simple monolayers to thick spheroid, organoid and tissue samples. The CX7 LZR also offers on-the-fly phenotyping that enables parallel image capture and analysis for multiplexed cytometry measurements in real time.
The Thermo Scientific CellInsight Pro High-Content Screening (HCS) Platform is an integrated LED system for all-around performance in screening and analysis. Equipped with best-in-class sCMOS camera featuring a near-perfect 95% quantum efficiency,  the CellInsight CX7 Pro platform enables you to select the right modes to read your sample both well-by-well and channel-by-channel. Use the entire fluorescence spectrum to optimize your assay and select either widefield or confocal optics for any channel. You also have 5-color brightfield options for colorimetric analysis of tissue sections.
A benchtop automated widefield screening system for researchers looking to increase scale and capacity in everyday use. With the compact and powerful CellInsight CX5 High-Content Screening Platform you can read and analyze a 96-well plate in under four minutes. Designed for researchers looking to increase their screening throughput, the platform offers common excitation wavelengths for fluorescence and white light illumination for colorimetric samples. The system is ready for full automation with 96-, 384-, or 1,536-well plates, or you can analyze your tissue sections one slide at a time to work up your next assay.
Best option for
Ideal for high-throughput screening and 3D organoid and spheroid research involving thick specimens.
Ideal for multiplex biology, including time-lapse and live-cell imaging.
Ideal for researchers needing optics that deliver optimal assay sensitivity.
Camera
sCMOS sCMOS CCD
Illumination
7 channels
7 channels
5 channels
Widefield
7-color laser excitation
7-color LED excitation
color LED excitation
Brightfield
4 specific LED + white light for higher contrast imaging (5-color total)
4 specific LED + white light for higher contrast imaging (5-color total)
White
Confocal
Nipkow spinning disk with 40 µm & 70 µm pinholes
Nipkow spinning disk with 40 µm & 70 µm pinholes
NA
Objectives
Olympus X-line objectives, magnifications 2x-60x
Focus
Laser and software
Laser and software
Software
Microplate types
6 to 1,536 wells
Onboard real-time analysis software
HCS Studio 5.0 Cell Analysis Software with Cell-painting Bioapplication
HCS Studio 4.0 Cell Analysis Software
EurekaScan Finder software
yes
yes
no
Z-stack acquisition
yes
yes
yes
Amira Software compatibility    
yes yes yes
Microscope slides
yes yes yes
Live-cell and time-lapse imaging
Optional on-stage incubator with kinetic scheduling of acquisition and analysis. CO2 and N2 for hypoxia conditions available.
Database
Store Express Image and Database Management Software Include: scalable SQL/Oracle database options available
Interested in the original version of the CX7?  See CX7 LZR (Cat. No. CX7A1110LZR) model.
For Research Use Only. Not for use in diagnostic procedures.