Temperature Controlled Chambers
Reach-in incubator for elevated temperature applications
Reach-In Refrigerated incubator broad temperature range covers incubation, testing and storage applications that require temperature control.
  • Reliable temperature control
  • Adjustable high and low temperature alarm prevents operator of deviations
  • Large chamber to accommodate large sample amounts 
To browse our complete portfolio or let one of our experts help you make a selection.
Contact us
Select from Reach-In Chambers
Reach-In Chamber
Reach-In Refrigerated Chamber
  reach-in-chamber reach-in-refrigerated-chamber
Size 29 cu. ft. 29 cu. ft.
Physical properties 29 cu.ft. Reach-in Chamber with heating 29 cu.ft. Reach-In Chamber with cooling and heating
Exterior (W x H x D) 965 x 2032 x 833mm
38.00 x 80.00 x 33.00 in.
965 x 2248 x 813mm
38.00 x 88.50 x 32.00 in.
Interior (W x H x D)
787 x 1524 x 686mm
31.00 x 60.00 x 27.00 in.
787 x 1524 x 686mm
31.00 x 60.00 x 27.00 in.
Net weight 227kg / 500 lbs 318kg / 700 lbs
Shipping weight 300kg / 660 lbs 386kg / 850 lbs
Temperature  
Temperature range 5°C above ambient to 60°C  0°C to 60°C
Controller, sensor Environ-Scan - PID, with RTD sensor Watlow Microprocessor - PID, with RTD sensor
Display Digital dual LED, 0.1°C steps readability & setability Digital dual LED, 0.1 C steps readability & setability
Temp. uniformity  ≤ ±0.4°C @ 30°C 
≤ ±0.3°C @ 37°C
≤ ±0.5°C @ 45°C
≤ ±0.3°C @ 20° to 37°C
Temp. stability @ 30° to 45°C @ 20° to 45°C
Electrical  
Voltage 115V, 50/60Hz, 9.0 Amps 3 Wire (including ground) 120V, 60Hz, 16.0 Amps 3 Wire
(including ground)
Plug Nema 5-15P NEMA 5-20P
Power switch 2-Pole
Interior convenience receptacles 230W max., for standard EU CEE7/7US Nema 5-15 plug Available as an option
Alarm contacts Temp. deviation and power failure; common, NO, NC; customer connections through RJ11 jack Temp. deviation and power failure; Common, NO, NC
Order No. 3960 3920
Reach-in chambers for elevated application temperatures 
Optional Cell Roll System
An optional cell roll system allows extensive production of monolayer cell cultures in standard roller culture vessels. This helps improved oxygenation and exposure of the cells to the media growth area. Combined uniform temperature control and continuous, gentile rotation provided by the cell roll system create the ideal conditions to help increase culture yields.
cell-roll-system
  • Cell roller up to 7 decks high with 5 positions per deck for a maximum total of 35 positions, or bottles.
  • Maximum capacity includes a Three-Tier Cell Roller Base (15 positions), four add-on tiers (20 positions), and a reinforced floor/ramp.
  • All position drive is standard. Adjustable speed contro lwith speeds of 0.125 to 6.25 RPM with ±1.0% accuracy, based on 110 mm bottles.