New: Ion AmpliSeq Transcriptome Mouse Gene Expression Kit
The Ion AmpliSeq Transcriptome Mouse Gene Expression Kit used with the Ion S5 System helps to enable fast and affordable gene expression analysis from samples limited in quantity. With simultaneous amplification of more than 20,000 RefSeq genes in a single tube, starting from as little as 10 ng of total RNA, this kit can be an important tool in oncology and translational research. The Ion AmpliSeq Transcriptome Mouse Gene Expression Kit offers:
  • Low sample input—as little as 10 ng of total RNA
  • Fast gene expression profiling with simple workflow—go from RNA to data in <2 days and only ~45 minutes of total hands-on time with the automated workflow of the Ion S5 and Ion Chef Systems
  • Streamlined data analysis—export data with Torrent Suite Software into existing analysis software
Ion AmpliSeq Transcriptome Mouse Gene Expression Kit flyer
Learn more about Targeted RNA Sequencing with Ion AmpliSeq RNA panels ›
View and order products
Correlation of DEGs between Ion AmpliSeq Mouse Gene Expression Kit data and whole transcriptome RNA-Seq data equals 0.943.
Ion AmpliSeq Transcriptome Human Gene Expression Kit
Transform your research by targeting >20,000 RefSeq transcripts, starting with as little as 10 ng of RNA input and a fast, FFPE-compatible workflow using the Ion AmpliSeq Transcriptome Human Gene Expression Kit and the Ion S5 series sequencing systems.
  • Achieve cost-effective transcriptome sequencing without waiting weeks to batch samples
  • Perform gene-level expression surveys with as little as 1 hour of hands-on time for library preparation, and go from RNA to gene expression levels in less than 2 days
  • Software tools included for fast and easy analysis of sequencing reads; use your existing downstream microarray or next-generation sequencing (NGS) pipelines for gene-level differential expression data analysis
  • Survey precious FFPE samples with as little as 10 ng of input RNA
Read the peer-reviewed publication:
Li et al (2015) Comprehensive evaluation of AmpliSeq transcriptome, a novel targeted whole transcriptome RNA sequencing methodology for global gene expression analysis. BMC Genomics 16:1069
Ion AmpliSeq Transcriptome Human Gene Expression Kit flyer
Read the Ion AmpliSeq Transcriptome app note to see a streamlined workflow for gene expression analysis
Read the Ion S5 application note to see how the Ion S5 System can be used for cancer genomics and transcriptomics research
View our ‘Webinar on Demand’ to learn how to apply a targeted NGS gene expression–based approach in biomarker discovery.
View Webinar on Demand
Looking for a gene expression signature in triple-negative breast cancer research samples using the Ion AmpliSeq Transcriptome Human Gene Expression kit
Is there only one way to do RNA-Seq library prep? Hear Milan Radovich, PhD, of Indiana University discuss his research on triple-negative breast cancer expression-based gene signatures.
Quantitative and differential gene expression analysis is fundamental to many biological research applications including gene regulation analysis, correlating gene expression with phenotypic information, and cellular pathway analysis. Conventional gene expression profiling methods include hybridization-based (microarray) and amplification-based (qRT-PCR) technologies. However, methods present certain limitations:
  • Despite requiring significant amounts of RNA, microarrays remain limited in their sensitivity and specificity
  • Although accepted as the gold standard for gene expression analysis, qRT-PCR is generally low-throughput, and substantial amounts of RNA are required if large numbers of genes are to be tested
As such, both technologies present workflow challenges when analyzing samples that are limited in quantity or quality, such as degraded RNA from FFPE tissues. Targeted sequencing using NGS and the Ion AmpliSeq Transcriptome Human Gene Expression Kit not only provides a representation of absolute expression, but also can identify and quantify low-abundance transcripts quickly and easily for both intact and degraded samples.
Improved gene-level transcript detection using the Ion AmpliSeq Transcriptome Human Gene Expression Kit and the Ion Proton System. A Venn diagram is shown demonstrating the concordance of genes identified by MAQC microarray data and the Ion AmpliSeq Transcriptome Human Gene Expression Kit.
Workflow for the Ion AmpliSeq Transcriptome Human or Mouse Gene Expression Kit
Amplify >20,000 targets in a single reaction
Products:
  • Ion AmpliSeq Transcriptome Human or Mouse Gene Expression Kit
Isothermal amplification with Ion Chef System
Products:
  • Ion 540 Kit-Chef
Learn more about Ion Chef System ›
Sequencing using Ion S5 System or Ion Proton System
Products:
  • Ion 540 Chip Kit
Learn more about Ion S5 System ›
  • Run AmpliSeqRNA plug-in to generate simple, easy-to-use read count data files
  • Perform simple statistical analysis or import files into existing microarray analysis pipelines
Learn more about Torrent Suite Software ›
Amplify >20,000 targets in a single reaction
Products:
  • Ion AmpliSeq Transcriptome Human or Mouse Gene Expression Kit
Isothermal amplification with Ion Chef System
Products:
  • Ion 540 Kit-Chef
Learn more about Ion Chef System ›
Sequencing using Ion S5 System or Ion Proton System
Products:
  • Ion 540 Chip Kit
Learn more about Ion S5 System ›
  • Run AmpliSeqRNA plug-in to generate simple, easy-to-use read count data files
  • Perform simple statistical analysis or import files into existing microarray analysis pipelines
Learn more about Torrent Suite Software ›
Ion S5 Systems
Adopting NGS in your lab is now simpler than ever before. The Ion S5 and Ion S5 XL next-generation sequencing systems enable the simplest targeted sequencing workflow for your lab with superior speed and affordability. Focus your research efforts on applications that have the potential to be impactful. The Ion S5 and Ion S5 XL Systems have a modular build, so your lab has the flexibility to do multiple targeted sequencing applications on a single system.
Learn more about the Ion S5 Systems ›
Visistat reference component