Search
Designed specifically to enhance each step of the SureTect PCR Assay Workflow, the SureTect PCR Workflow tools enable ease of-handling, reduced hands-on time and operator error, providing a superior user handling experience in your rapid pathogen detection.
In your busy Salmonella and Cronobacter testing laboratory workflows, there is no time for error. A common mistake, particularly in high throughout labs, is the accidental omission or duplication of adding a selective supplement during sample preparation. For clearer selective enrichment of food and environmental samples, the Thermo Scientific Oxoid PrecisBlue Supplement has been specifically designed to reduce workflow errors. The addition of an inert blue dye provides a clear visual reference, enabling easy identification of supplemented food and environmental samples.
Easy-to-use tools to simplify your rapid pathogen detection workflow: designed specifically to enhance each step of the SureTect PCR Assay workflow, Thermo Scientific SureTect Tools enable ease of handling, reducing hands-on time, risk of contamination and human error, providing a superior user handling experience.
| Product | Description | Cat. No. | |
|---|---|---|---|
![]() |
PCR Strip Cutting Tool |
For cutting Lysis and PCR Assay Strips when fewer than 8 tubes are needed |
A55329 |
![]() |
PCR Uncapping Tool for Lysis Tubes |
For removal of Domed Strip Caps from Lysis Strips |
A55328 |
![]() |
PCR Capping Tool |
For capping Lysis and PCR Assay Plate strips |
A55327 |
![]() |
MicroAmp Adhesive Film Applicator |
Pack of 5; for ease of applying the Pierceable Film to Lysis Plate strips |
4333183 |
PCR Strip Cutting Tool:
Capping Tool with Domed Lysis Tube Caps:
Uncapping Tool with Domed Lysis Tube Caps:
SureTect PCR Workflow video:
| Product | Description | Cat. No. |
|---|---|---|
SimpliAmp Strip Plate Tray |
For compatibility and ease of loading SureTect Lysis Tubes into the SimpliAmp Thermal Cycler |
|
QuantStudio 5 Strip Plate Tray |
For compatibility and ease of loading SureTect PCR Assay Tubes into the QuantStudio 5 PCR instrument |
A55332 |
| QuantStudio 5 Tray Cover (optional) | To aid in vortexing and spinning the PCR tubes. | A55972 |
SimpliAmp Strip Plate Tray Cover |
Pack of 10 for covering and securing the Lysis Tubes before pipetting |
A55971 |
96-well PCR Base |
96-well base for Lysis and PCR Assay Strip Plates. Use in Lysis and PCR steps | A55330 |
Each SureTect PCR Assay comes with all the reagents and PCR accessories needed to set up your SureTect PCR Assay tests to run on the QuantStudio 5 Food Safety PCR System. Additional SureTect Accessories are available to order separately, for example, domed lysis tube caps for use if preferred in place of the pierceable lysis seals.
| Product | Description | Cat. No. |
|---|---|---|
96-well pierceable lysis seal |
Pack of 100 seals
|
A55331 |
Pack of 120 strips: Optional domed caps that can be used in place of pierceable seals for the Lysis Tubes |
A56895 | |
For balancing of 7500FAST plates |
AB2696 | |
Ultra clear qPCR caps |
Additional PCR caps, if required |
AB0866 |
For Research Use Only. Not for use in diagnostic procedures.